Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00459980_m1
          See other CD209E GE Assays ›
          Gene Symbol:
          Cd209e
          Gene Name:
          CD209e antigen
          Gene Aliases:
          SIGNR4, mSIGNR4
          Chromosome Location:
          Chr.8: 3847973 - 3854286 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00459980_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd209e
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_130905.2 NP_570975.1 5 - 6 507 60
          GenBank mRNA AF373412.1 - 5 - 6 507 60
          AK040917.1 - 5 - 6 529 60
          BC107188.2 - 5 - 6 507 40050810 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001622.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cd209e

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          endocytosis
          biological_process
          mannose binding
          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1960376 NM_130905.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1960376 AF373412.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1960376 AK040917.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 554393 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 324359 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 633286 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 6349 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 816762 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 87589 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P146304 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1960376 NM_130905.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1960376 AF373412.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1960376 AK040917.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P495311 NM_130905.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P495311 AF373412.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P495311 AK040917.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P495311 BC107188.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615616 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615611 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615608 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615623 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615627 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615614 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615630 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615641 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615639 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615642 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615629 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615640 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615636 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615601 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615605 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615619 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615632 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06408316 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P146304 - -
          Whole Mouse Genome G4122A A_51_P495311 NM_130905.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P495311 AF373412.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P495311 AK040917.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P495311 BC107188.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135177 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P146304 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06673349 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06673359 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06673358 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06978095 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06673362 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07033363 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06673335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06979622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06979717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06978095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07446967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07194856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07283950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06843002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06673357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615581 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07025886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07234257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06843000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615593 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10576804 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576806 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576805 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576800 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576803 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576801 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576802 - -
          Affymetrix HT MG-430 PM Array Plate 1420582_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10576799 - -
          Mouse Genome 430 2.0 Array 1420582_at - -
          Mouse Genome 430 2.0 Array 1420582_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 170780_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 170780_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0800013334.mm.1 NM_130905.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013334.mm.1 NM_130905.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013334.mm.1 TC0800001622.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800013335.mm.1 NM_130905.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013335.mm.1 NM_130905.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013335.mm.1 TC0800001622.mm.1 MTA TC
          Mouse Expression Set 430 1420582_PM_at - -
          Mouse Expression Set 430 1420582_at - -
          Mouse Genome 430A 2.0 Array 1420582_PM_at - -
          Mouse Genome 430A 2.0 Array 1420582_at - -
          Affymetrix HT Mouse Genome 430A Array 1420582_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420582_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 564529 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4280044 - -
          Illumina mouse-6 v1.0 expression beadchip 4280044 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004280044 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2665757 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2665757 - -
          mouse-6 v1.1 (Illumina) scl35150.6.1_245-S - -
          Illumina Sentrix mouse-6.v1 bead array 4280044 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl35150.6.1_245-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4280044 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2665757 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2665757 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline