Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00460844_m1
          See other MGL2 GE Assays ›
          Gene Symbol:
          Mgl2
          Gene Name:
          macrophage galactose N-acetyl-galactosamine specific lectin 2
          Gene Aliases:
          CD301b
          Chromosome Location:
          Chr.11: 70130125 - 70137542 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00460844_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mgl2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145137.2 NP_660119.1 2 - 3 225 70
          GenBank mRNA AK085751.1 - 1 - 2 85 70
          AY103461.1 - 2 - 3 162 70
          BC106833.1 - 1 - 2 151 40044847 70
          BC110475.2 - 1 - 2 151 40045765 70
          BC111520.1 - 1 - 2 151 40042306 70
          BY198732.1 - 2 - 3 227 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000941.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mgl2

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Process(es)

          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2128499 EU010011.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2128499 AY103461.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2128499 BY544307.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2128499 NM_145137.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 723891 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 776963 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 14738 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 176527 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P138920 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2128499 EU010011.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2128499 AY103461.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2128499 BY544307.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2128499 NM_145137.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899139 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899133 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899126 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899127 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899143 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899121 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899145 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899124 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899125 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07387735 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P138920 - -
          Whole Mouse Genome G4122A A_52_P414065 EU010011.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P414065 BC106833.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P414065 BC110475.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P414065 AY103461.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P414065 BC111520.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112801 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P138920 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09430420 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09430415 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09430408 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09430405 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09430406 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09430425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09430420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09447009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09658343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09430409 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09561636 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09538411 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09430415 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09502145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09430425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899135 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09502132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09561631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09409224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09538409 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09342431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09385842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899113 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09430406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899141 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10377777 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377776 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377780 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377779 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377778 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377775 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377781 - -
          Affymetrix HT MG-430 PM Array Plate 1438467_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10377774 - -
          Mouse Genome 430 2.0 Array 1438467_at - -
          Mouse Genome 430 2.0 Array 1438467_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 216864_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 216864_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100008224.mm.1 NM_145137.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008224.mm.1 NM_145137.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008224.mm.1 TC1100000941.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100008230.mm.1 NM_145137.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008230.mm.1 NM_145137.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008230.mm.1 TC1100000941.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100008231.mm.1 NM_145137.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008231.mm.1 NM_145137.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008231.mm.1 TC1100000941.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100008232.mm.1 NM_145137.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008232.mm.1 NM_145137.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008232.mm.1 TC1100000941.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100008227.mm.1 NM_145137.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008227.mm.1 NM_145137.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008227.mm.1 TC1100000941.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100008221.mm.1 NM_145137.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008221.mm.1 NM_145137.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008221.mm.1 TC1100000941.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100008226.mm.1 NM_145137.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008226.mm.1 NM_145137.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008226.mm.1 TC1100000941.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100008218.mm.1 NM_145137.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008218.mm.1 NM_145137.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008218.mm.1 TC1100000941.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100008222.mm.1 NM_145137.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008222.mm.1 NM_145137.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008222.mm.1 TC1100000941.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100008223.mm.1 NM_145137.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008223.mm.1 NM_145137.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008223.mm.1 TC1100000941.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100008228.mm.1 NM_145137.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008228.mm.1 NM_145137.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008228.mm.1 TC1100000941.mm.1 MTA TC
          Mouse Expression Set 430 1438467_at - -
          Mouse Expression Set 430 1438467_PM_at - -
          Murine Genome U74v2 Set 133755_PM_at - -
          Murine Genome U74v2 Set 133755_at - -
          Mouse Genome 430A 2.0 Array 1438467_at - -
          Mouse Genome 430A 2.0 Array 1438467_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1438467_at - -
          Affymetrix HT Mouse Genome 430A Array 1438467_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 668380 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5690441 - -
          Illumina mouse-6 v1.0 expression beadchip 6180008 - -
          Illumina mouse-6 v1.0 expression beadchip 5690441 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006180008 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005690441 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2707100 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2707319 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2707100 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2707319 - -
          mouse-6 v1.1 (Illumina) scl41343.8_1-S - -
          mouse-6 v1.1 (Illumina) scl0001401.1_29-S - -
          Illumina Sentrix mouse-6.v1 bead array 6180008 - -
          Illumina Sentrix mouse-6.v1 bead array 5690441 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0001401.1_29-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl41343.8_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6180008 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5690441 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2707099 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2707319 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2707099 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline