Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00461780_m1
          See other JCHAIN GE Assays ›
          Gene Symbol:
          Jchain
          Gene Name:
          immunoglobulin joining chain
          Gene Aliases:
          9530090F24Rik, AI323815, Igj, Jch
          Chromosome Location:
          Chr.5: 88519809 - 88527897 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00461780_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Jchain
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_152839.3 NP_690052.2 2 - 3 306 96
          GenBank mRNA AB644392.1 - 2 - 3 188 96
          AK078865.1 - 2 - 3 192 96
          AK160344.1 - 2 - 3 258 96
          BC006026.1 - 2 - 3 210 3491646 96
          J00544.1 - 2 - 3 193 96
          M90766.1 - 2 - 3 209 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002712.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Jchain

          Panther Classification:

          Molecular Function -

          defense/immunity protein immunoglobulin

          Gene Ontology Categories:

          Function(s) Process(es)

          adaptive immune response
          glomerular filtration
          humoral immune response
          antibacterial humoral response
          positive regulation of protein oligomerization
          innate immune response
          positive regulation of respiratory burst
          single-stranded DNA binding
          antigen binding
          IgA binding
          phosphatidylcholine binding
          immunoglobulin receptor binding
          protein homodimerization activity
          peptidoglycan binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150710 BC006026.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150710 AK078865.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150710 NM_152839.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150710 BC006026.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150710 AK078865.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150710 NM_152839.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 910531 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 748102 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 20362 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 541396 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 855826 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 94823 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 147293 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150710 BC006026.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150710 AK078865.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150710 NM_152839.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150710 BC006026.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150710 AK078865.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150710 NM_152839.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P150705 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P150710 BC006026.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P150710 AK078865.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P150710 NM_152839.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P150710 BC006026.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P150710 AK078865.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P150710 NM_152839.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23872711 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23872701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23872699 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23872702 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23872698 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23872704 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23872707 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23872709 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23872713 - -
          Whole Mouse Genome G4122A A_51_P150710 BC006026.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150710 AK078865.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150710 NM_152839.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150710 BC006026.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150710 AK078865.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150710 NM_152839.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150705 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132372 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04117909 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04117906 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04117905 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04117904 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04117908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23872709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872718 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872701 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04849864 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872704 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872703 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872724 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872712 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04335844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04216178 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872714 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872702 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872725 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04266288 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872715 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872722 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04820090 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872698 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872719 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23872707 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10531129 - -
          Affymetrix Mouse Gene 1.0 ST Array 10531127 - -
          Affymetrix Mouse Gene 1.0 ST Array 10531128 - -
          Affymetrix Mouse Gene 1.0 ST Array 10531131 - -
          Affymetrix Mouse Gene 1.0 ST Array 10531132 - -
          Affymetrix Mouse Gene 1.0 ST Array 10531130 - -
          Affymetrix HT MG-430 PM Array Plate 1424305_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10531126 - -
          Mouse Genome 430 2.0 Array 1424305_PM_at - -
          Mouse Genome 430 2.0 Array 1424305_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16069_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16069_at - -
          Affymetrix Mouse Clariom S Array PSR0500023903.mm.1 TC0500002712.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500023903.mm.1 NM_152839.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023903.mm.1 NM_152839.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023910.mm.1 TC0500002712.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500023910.mm.1 NM_152839.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023910.mm.1 NM_152839.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023906.mm.1 TC0500002712.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500023906.mm.1 NM_152839.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023906.mm.1 NM_152839.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023911.mm.1 TC0500002712.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500023911.mm.1 NM_152839.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023911.mm.1 NM_152839.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023908.mm.1 TC0500002712.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500023908.mm.1 NM_152839.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023908.mm.1 NM_152839.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023904.mm.1 TC0500002712.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500023904.mm.1 NM_152839.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023904.mm.1 NM_152839.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023905.mm.1 TC0500002712.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500023905.mm.1 NM_152839.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023905.mm.1 NM_152839.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023907.mm.1 TC0500002712.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500023907.mm.1 NM_152839.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023907.mm.1 NM_152839.3 Refseq
          Mouse Expression Set 430 1424305_at - -
          Mouse Expression Set 430 1424305_PM_at - -
          Murine Genome U74v2 Set 102372_PM_at - -
          Murine Genome U74v2 Set 102372_at - -
          Mouse Genome 430A 2.0 Array 1424305_at - -
          Mouse Genome 430A 2.0 Array 1424305_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424305_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424305_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 749478 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1170541 - -
          Illumina mouse-6 v1.0 expression beadchip 1170541 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001170541 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1221256 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1221256 - -
          mouse-6 v1.1 (Illumina) scl0016069.1_4-S - -
          Illumina Sentrix mouse-6.v1 bead array 1170541 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0016069.1_4-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1170541 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1221256 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1221256 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline