Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00462019_m1
          See other LRRC25 GE Assays ›
          Gene Symbol:
          Lrrc25
          Gene Name:
          leucine rich repeat containing 25
          Gene Aliases:
          Mapa
          Chromosome Location:
          Chr.8: 70616844 - 70620850 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00462019_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lrrc25
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153074.3 NP_694714.1 2 - 3 986 56
          GenBank mRNA AJ422149.1 - 1 - 2 803 56
          AK080022.1 - 2 - 3 987 56
          AK171924.1 - 2 - 3 976 56
          BC137696.1 - 2 - 3 946 8860715 56

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800000758.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lrrc25

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P108108 AK171924.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P108108 NM_153074.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P108108 AK080022.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P108108 AK171924.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P108108 NM_153074.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P108108 AK080022.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 9092 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 287827 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 294672 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163732 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P108108 AK171924.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P108108 NM_153074.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P108108 AK080022.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P108108 AK171924.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P108108 NM_153074.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P108108 AK080022.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P108108 AK171924.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P108108 NM_153074.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P108108 AK080022.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P108108 AK171924.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P108108 NM_153074.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P108108 AK080022.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953048 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953046 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953038 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953040 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06549135 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163732 - -
          Whole Mouse Genome G4122A A_51_P108108 AK171924.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P108108 NM_153074.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P108108 AK080022.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P108108 AK171924.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P108108 NM_153074.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P108108 AK080022.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101571 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06922000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06894473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07066550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06922013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06846490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06894474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07269134 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953031 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06922012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07207659 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10572446 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572448 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572447 - -
          Affymetrix Mouse Gene 1.1 ST Array 10572445 - -
          Affymetrix Mouse Clariom S Array PSR0800005376.mm.1 NM_153074.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005376.mm.1 NM_153074.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005376.mm.1 TC0800000758.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800005375.mm.1 NM_153074.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005375.mm.1 NM_153074.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005375.mm.1 TC0800000758.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 431994 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3360592 - -
          Illumina mouse-6 v1.0 expression beadchip 3360592 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003360592 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2715800 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2715800 - -
          mouse-6 v1.1 (Illumina) scl0211228.2_29-S - -
          Illumina Sentrix mouse-6.v1 bead array 3360592 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0211228.2_29-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3360592 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2715800 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2715800 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0