Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00467065_m1
          See other CRK GE Assays ›
          Gene Symbol:
          Crk
          Gene Name:
          v-crk sarcoma virus CT10 oncogene homolog (avian)
          Gene Aliases:
          Crk-I, Crk-II, Crk-III, Crk3, CrkIII, Crko, c-Crk, p38
          Chromosome Location:
          Chr.11: 75679248 - 75708428 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00467065_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Crk
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001277219.1 NP_001264148.1 1 - 2 410 137
          NM_133656.5 NP_598417.2 1 - 2 410 137
          GenBank mRNA AF239673.1 - 1 - 2 280 137
          AK028179.1 - 1 - 2 387 137
          AK028488.1 - 1 - 2 360 137
          AK036905.1 - 1 - 2 361 137
          AK052795.1 - 1 - 2 369 137
          AK076097.1 - 1 - 2 105 137
          AK088519.1 - 1 - 2 381 137
          AK139089.1 - 1 - 2 366 137
          AK152549.1 - 1 - 2 384 137
          AK154987.1 - 1 - 2 395 137
          AK161356.1 - 1 - 2 360 137
          AK161761.1 - 1 - 2 352 137
          BC012216.1 - 1 - 2 346 3992341 137
          BC031149.1 - 1 - 2 343 4975654 137
          S72408.1 - 1 - 2 335 137

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001078.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Crk

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of signal transduction
          regulation of actin cytoskeleton organization
          regulation of Rac protein signal transduction
          regulation of GTPase activity
          ephrin receptor signaling pathway
          SH3/SH2 adaptor activity
          protein binding
          protein domain specific binding
          protein binding, bridging
          SH2 domain binding
          protein phosphorylated amino acid binding
          ephrin receptor binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100009892.mm.1 NM_001277219.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009892.mm.1 NM_133656.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009892.mm.1 TC1100001078.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100009886.mm.1 NM_001277219.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009886.mm.1 NM_001277221.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009886.mm.1 NM_133656.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009886.mm.1 TC1100001078.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100009885.mm.1 NM_001277219.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009885.mm.1 NM_001277221.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009885.mm.1 NM_133656.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009885.mm.1 TC1100001078.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100009890.mm.1 NM_001277219.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009890.mm.1 NM_001277221.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009890.mm.1 NM_133656.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009890.mm.1 TC1100001078.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100009891.mm.1 NM_001277219.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009891.mm.1 NM_001277221.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009891.mm.1 NM_133656.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009891.mm.1 TC1100001078.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 434229 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline