Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00468801_m1
          See other H2-OB GE Assays ›
          Gene Symbol:
          H2-Ob
          Gene Name:
          histocompatibility 2, O region beta locus
          Gene Aliases:
          A-beta-2, A-beta2, H-2Ab, H-2I, H-2Ob, H2-Ab, H2-Ab2, H2-IAb2, Ob
          Chromosome Location:
          Chr.17: 34238895 - 34245908 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00468801_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: H2-Ob
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010389.3 NP_034519.2 1 - 2 162 68
          GenBank mRNA AK155144.1 - 1 - 2 112 68
          AK155650.1 - 1 - 2 162 68
          AK172191.1 - 1 - 2 167 68
          AK172273.1 - 1 - 2 137 68
          AK172299.1 - 1 - 2 137 68
          BC127027.1 - 1 - 2 95 40044586 68
          BC127028.1 - 1 - 2 95 40044587 68
          KJ650230.1 - 1 - 2 95 68
          KJ663722.1 - 1 - 2 95 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000625.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene H2-Ob

          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of antigen processing and presentation of peptide antigen via MHC class II
          MHC class II protein complex binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P130916 NM_010389.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2082929 AK155650.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2082929 NM_010389.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 313405 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 229028 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 666914 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 648094 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P130916 NM_010389.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2082929 AK155650.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2082929 NM_010389.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2057931 AK080011.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P679470 AK053233.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P679470 NM_010389.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152821 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152826 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152816 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152824 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152823 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152817 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152825 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152813 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152814 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152818 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152828 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08764917 - -
          Whole Mouse Genome G4122A A_52_P679470 AK053233.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P679470 NM_010389.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126717 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130916 NM_010389.3 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08764917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152819 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152821 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152817 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152818 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152816 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152826 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152833 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152824 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152835 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152812 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152823 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152830 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152813 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152838 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152822 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152828 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10444289 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444287 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444285 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444288 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444286 - -
          Affymetrix HT MG-430 PM Array Plate 1422201_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1440837_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10444284 - -
          Mouse Genome 430 2.0 Array 1440837_at - -
          Mouse Genome 430 2.0 Array 1422201_PM_at - -
          Mouse Genome 430 2.0 Array 1422201_at - -
          Mouse Genome 430 2.0 Array 1440837_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15002_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15002_at - -
          Affymetrix Mouse Clariom S Array PSR1700006005.mm.1 NM_010389.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006005.mm.1 NM_010389.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006005.mm.1 TC1700000625.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700006006.mm.1 NM_010389.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006006.mm.1 NM_010389.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006006.mm.1 TC1700000625.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700006010.mm.1 NM_010389.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006010.mm.1 NM_010389.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006010.mm.1 TC1700000625.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700006012.mm.1 NM_010389.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006012.mm.1 NM_010389.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006012.mm.1 TC1700000625.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700006004.mm.1 NM_010389.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006004.mm.1 NM_010389.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006004.mm.1 TC1700000625.mm.1 MTA TC
          Mouse Expression Set 430 1422201_at - -
          Mouse Expression Set 430 1422201_PM_at - -
          Mouse Expression Set 430 1440837_at - -
          Mouse Expression Set 430 1440837_PM_at - -
          Murine Genome U74v2 Set 104429_at - -
          Murine Genome U74v2 Set 104429_PM_at - -
          Mouse Genome 430A 2.0 Array 1422201_PM_at - -
          Mouse Genome 430A 2.0 Array 1422201_at - -
          Affymetrix HT Mouse Genome 430A Array 1422201_at - -
          Affymetrix HT Mouse Genome 430A Array 1422201_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440837_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440837_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6980059 - -
          Illumina mouse-6 v1.0 expression beadchip 102690047 - -
          Illumina mouse-6 v1.0 expression beadchip 6980059 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006980059 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2670690 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2670690 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2502380 - -
          mouse-6 v1.1 (Illumina) scl50826.4.1_157-S - -
          mouse-6 v1.1 (Illumina) scl8544.1.1_308-S - -
          Illumina Sentrix mouse-6.v1 bead array 6980059 - -
          Illumina Sentrix mouse-6.v1 bead array 102690047 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50826.4.1_157-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102690047 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6980059 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2670690 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2670690 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline