Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00469962_m1
          See other RGL3 GE Assays ›
          Gene Symbol:
          Rgl3
          Gene Name:
          ral guanine nucleotide dissociation stimulator-like 3
          Gene Aliases:
          1300003D20Rik, mFLJ00153
          Chromosome Location:
          Chr.9: 21971179 - 21989465 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00469962_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rgl3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023622.4 NP_076111.2 9 - 10 1249 80
          GenBank mRNA AF237669.1 - 9 - 10 1208 80
          AF239661.1 - 9 - 10 1250 80
          AK004876.1 - 8 - 9 982 80
          AK053550.1 - 9 - 10 1236 80
          AK131140.1 - 755 80
          AK134654.1 - 1231 80
          AK165746.1 - 9 - 10 1243 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900001890.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rgl3

          Panther Classification:

          Molecular Function -

          guanyl-nucleotide exchange factor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          small GTPase mediated signal transduction
          positive regulation of GTPase activity
          guanyl-nucleotide exchange factor activity
          protein binding
          Ral guanyl-nucleotide exchange factor activity
          Ras GTPase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26349892 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2086825 AK131140.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2086825 AK134654.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2086825 AF239661.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2086825 NM_023622.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 451665 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 425932 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 455637 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 952934 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 821954 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 860569 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 823994 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 484107 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 751697 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 111602 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 213011 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 673744 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 74623 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 82051 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P130692 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P127761 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2086825 AK131140.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2086825 AK134654.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2086825 AF239661.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2086825 NM_023622.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26349895 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26349902 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26349890 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26349896 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26349888 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26349899 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26349881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26349905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26349900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26349886 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26349885 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26349893 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26349898 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26349894 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P127761 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06720119 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P130692 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06720153 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06720164 - -
          Whole Mouse Genome G4122A A_52_P440599 AK165023.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P440599 AK131140.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P440599 AK053550.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P440599 AK134654.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P440599 AF239661.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139034 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133292 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P130692 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P127761 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07903848 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07903826 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07903833 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07903859 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07903821 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07903823 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26349892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26349829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26349896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07728472 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26349833 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26349877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08046990 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26349842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26349890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26349885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07903845 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07916824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07643425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26349881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26349905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26349886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07583711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26349828 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07903826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07831701 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26349863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26349829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26349836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26349874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26349868 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07837781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07884440 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26349893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26349830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08055994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26349831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26349835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26349827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26349894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26349899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26349880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26349882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26349840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26349876 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26349900 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10591679 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591676 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591689 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591685 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591688 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591673 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591681 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591670 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591677 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591684 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591678 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591680 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591690 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591687 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591672 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591674 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591669 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591671 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591675 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591686 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591682 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591683 - -
          Affymetrix HT MG-430 PM Array Plate 1431829_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10591668 - -
          Mouse Genome 430 2.0 Array 1431829_a_at - -
          Mouse Genome 430 2.0 Array 1431829_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71746_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71746_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900015934.mm.1 NM_023622.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900015934.mm.1 NM_023622.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900015934.mm.1 TC0900001890.mm.1 MTA TC
          Mouse Expression Set 430 1431829_PM_a_at - -
          Mouse Expression Set 430 1431829_a_at - -
          Mouse Genome 430A 2.0 Array 1431829_a_at - -
          Mouse Genome 430A 2.0 Array 1431829_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1431829_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1431829_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 928493 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6550025 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006550025 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1239492 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1239492 - -
          mouse-6 v1.1 (Illumina) scl36127.13.1_2-S - -
          Illumina Sentrix mouse-6.v1 bead array 6550025 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl36127.13.1_2-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6550025 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2991779 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1239492 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1239492 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2991779 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline