Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00471689_g1
          See other 1700067P10RIK GE Assays ›
          Gene Symbol:
          1700067P10Rik
          Gene Name:
          RIKEN cDNA 1700067P10 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 48089318 - 48090920 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00471689_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700067P10Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026625.2 NP_080901.2 2 - 3 112 67
          GenBank mRNA AK018887.1 - 2 - 3 101 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000930.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 1700067P10Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31215888 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31215872 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31215883 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31215881 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31215874 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31215873 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31215880 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31215882 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31215871 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31215889 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31215887 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P271268 AK018887.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P271268 AK018887.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 622548 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P150105 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P271268 AK018887.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P271268 AK018887.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31215827 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31215840 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31215815 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31215839 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31215835 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31215816 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31215811 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31215823 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31215822 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31215831 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31215825 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31215842 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31215828 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31215845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31215833 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31215837 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31215847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31215830 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31215849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31215819 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31215813 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P150105 - -
          Whole Mouse Genome G4122A A_51_P271268 AK018887.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P271268 AK018887.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118569 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P150105 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13465382 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13262192 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13465377 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31215871 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31215883 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31215887 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31215880 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31215874 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31215872 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31215873 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31215889 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31215882 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31215888 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31215881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215867 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13465375 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215885 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13398506 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13199190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215849 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215788 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215877 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13149367 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215856 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215801 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13516893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215871 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215802 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215881 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215809 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13398502 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13160323 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13359276 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215883 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215816 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215819 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215865 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215863 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13465379 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215903 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215797 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215875 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215895 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215835 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215811 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13465382 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215860 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13414712 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13398503 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215888 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215898 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13444350 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215786 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215872 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215821 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215827 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13199182 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215844 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215824 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215808 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13516892 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215814 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13262194 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13262196 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13199188 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10445745 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445744 - -
          Affymetrix HT MG-430 PM Array Plate 1420724_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10445743 - -
          Mouse Genome 430 2.0 Array 1420724_PM_at - -
          Mouse Genome 430 2.0 Array 1420724_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68224_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68224_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700008965.mm.1 NM_026625.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008965.mm.1 NM_026625.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008965.mm.1 TC1700000930.mm.1 MTA TC
          Mouse Expression Set 430 1420724_at - -
          Mouse Expression Set 430 1420724_PM_at - -
          Mouse Genome 430A 2.0 Array 1420724_PM_at - -
          Mouse Genome 430A 2.0 Array 1420724_at - -
          Affymetrix HT Mouse Genome 430A Array 1420724_at - -
          Affymetrix HT Mouse Genome 430A Array 1420724_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 402768 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6380538 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006380538 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2616500 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2616500 - -
          mouse-6 v1.1 (Illumina) scl50641.3.1_40-S - -
          Illumina Sentrix mouse-6.v1 bead array 6380538 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50641.3.1_40-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6380538 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2616500 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2616500 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline