Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00471820_m1
          See other PPP1R14A GE Assays ›
          Gene Symbol:
          Ppp1r14a
          Gene Name:
          protein phosphatase 1, regulatory (inhibitor) subunit 14A
          Gene Aliases:
          1110001M11Rik, Cpi17
          Chromosome Location:
          Chr.7: 29289320 - 29293390 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00471820_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppp1r14a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026731.3 NP_081007.2 3 - 4 329 65
          GenBank mRNA AK003249.1 - 3 - 4 327 65
          AY050672.1 - 3 - 4 328 65
          BC010832.1 - 3 - 4 329 4216018 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000527.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ppp1r14a

          Panther Classification:

          Molecular Function -

          phosphatase modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of phosphorylation
          protein phosphatase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25026384 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25026385 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P432460 AK003249.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P432460 NM_026731.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P432460 AF352573.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P432460 AY050672.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P432460 BC010832.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P432460 AK003249.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P432460 NM_026731.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P432460 AF352573.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P432460 AY050672.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P432460 BC010832.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 564644 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 696065 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 421238 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 163470 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121725 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P432460 AK003249.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P432460 NM_026731.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P432460 AF352573.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P432460 AY050672.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P432460 BC010832.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P432460 AK003249.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P432460 NM_026731.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P432460 AF352573.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P432460 AY050672.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P432460 BC010832.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25026396 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25026390 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25026387 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25026395 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25026399 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25026391 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25026384 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25026389 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25026394 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25026392 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25026385 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25026393 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25026400 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P121725 - -
          Whole Mouse Genome G4122A A_51_P432460 AK003249.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P432460 NM_026731.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P432460 AF352573.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P432460 AY050672.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P432460 BC010832.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P432460 AK003249.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P432460 NM_026731.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P432460 AF352573.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P432460 AY050672.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P432460 BC010832.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107927 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P121725 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25026385 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25026384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026333 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25026409 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25026387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25026393 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25026379 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25026404 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25026382 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25026392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25026391 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25026390 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25026411 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25026403 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06034620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25026373 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25026400 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25026412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25026396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06015835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25026376 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25026369 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06034625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06660229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25026407 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06616419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06162692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25026368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25026399 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25026406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25026371 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25026395 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25026413 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10551739 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551740 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551737 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551738 - -
          Affymetrix HT MG-430 PM Array Plate 1418086_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10551736 - -
          Mouse Genome 430 2.0 Array 1418086_PM_at - -
          Mouse Genome 430 2.0 Array 1418086_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68458_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68458_at - -
          Affymetrix Mouse Clariom S Array PSR0700004491.mm.1 TC0700000527.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700004491.mm.1 NM_026731.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004491.mm.1 NM_026731.3 Refseq
          Mouse Expression Set 430 1418086_PM_at - -
          Mouse Expression Set 430 1418086_at - -
          Mouse Genome 430A 2.0 Array 1418086_PM_at - -
          Mouse Genome 430A 2.0 Array 1418086_at - -
          Affymetrix HT Mouse Genome 430A Array 1418086_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418086_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 773044 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 870114 - -
          MouseWG-6 v2 Expression BeadChip 4570215 - -
          Illumina mouse-6 v1.0 expression beadchip 4570215 - -
          Illumina mouse-6 v1.0 expression beadchip 870114 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004570215 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000870114 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2659824 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2729363 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2659824 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2729363 - -
          mouse-6 v1.1 (Illumina) scl00061.1_10-S - -
          mouse-6 v1.1 (Illumina) scl32843.5.1_71-S - -
          Illumina Sentrix mouse-6.v1 bead array 870114 - -
          Illumina Sentrix mouse-6.v1 bead array 4570215 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00061.1_10-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32843.5.1_71-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4570215 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 870114 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2729363 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2659824 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2659824 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2729363 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl00061.1_10-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl32843.5.1_71-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline