Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00474243_s1
          See other PCDHB4 GE Assays ›
          Gene Symbol:
          Pcdhb4
          Gene Name:
          protocadherin beta 4
          Gene Aliases:
          Pcdhb5A, PcdhbD
          Chromosome Location:
          Chr.18: 37307455 - 37311173 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00474243_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pcdhb4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053129.3 NP_444359.3 1 2494 82
          GenBank mRNA AK029277.1 - 1 2486 82
          AK036169.1 - 1 2495 82
          AK143280.1 - 2 1821 82
          AK160730.1 - 1 2495 82
          AY013786.1 - 1 2310 82
          BC053098.1 - 1 2467 6401988 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800000358.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pcdhb4

          Panther Classification:

          Molecular Function -

          cadherin

          Gene Ontology Categories:

          Process(es)

          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P529195 BC053098.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P529195 AK143280.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P529195 AK160730.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P529195 AK029277.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P529195 AK036169.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P529195 BC053098.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P529195 AK143280.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P529195 AK160730.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P529195 AK029277.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P529195 AK036169.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 855433 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 26106 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139148 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P529195 BC053098.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P529195 AK143280.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P529195 AK160730.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P529195 AK029277.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P529195 AK036169.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P529195 BC053098.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P529195 AK143280.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P529195 AK160730.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P529195 AK029277.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P529195 AK036169.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627726 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627741 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627729 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627742 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627737 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627750 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627749 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627733 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627731 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627745 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627739 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627751 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627736 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627743 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139148 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08967966 - -
          Whole Mouse Genome G4122A A_52_P529195 BC053098.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P529195 AK143280.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P529195 AK160730.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P529195 AK029277.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P529195 AK036169.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P529195 BC053098.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P529195 AK143280.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P529195 AK160730.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P529195 AK029277.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P529195 AK036169.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136738 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139148 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13965789 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13965748 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13677888 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13965775 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13965784 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13965777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627720 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13677900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627773 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627742 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627775 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627750 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13677891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13965750 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627737 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627780 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13677888 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627741 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13965786 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627777 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627736 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627743 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627754 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627776 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627761 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627758 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627774 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13892263 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627748 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627771 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627751 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627739 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10455063 - -
          Affymetrix Mouse Gene 1.0 ST Array 10455064 - -
          Affymetrix Mouse Gene 1.0 ST Array 10455062 - -
          Affymetrix HT MG-430 PM Array Plate 1422951_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1440632_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10455061 - -
          Mouse Genome 430 2.0 Array 1422951_PM_at - -
          Mouse Genome 430 2.0 Array 1440632_at - -
          Mouse Genome 430 2.0 Array 1440632_PM_at - -
          Mouse Genome 430 2.0 Array 1422951_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 93875_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 93875_at - -
          Affymetrix Mouse Clariom S Array PSR1800002819.mm.1 NM_053129.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002819.mm.1 NM_053129.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002819.mm.1 TC1800000358.mm.1 MTA TC
          Mouse Expression Set 430 1422951_PM_at - -
          Mouse Expression Set 430 1422951_at - -
          Mouse Expression Set 430 1440632_PM_at - -
          Mouse Expression Set 430 1440632_at - -
          Mouse Genome 430A 2.0 Array 1422951_at - -
          Mouse Genome 430A 2.0 Array 1422951_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422951_at - -
          Affymetrix HT Mouse Genome 430A Array 1422951_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440632_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440632_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 432260 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 670184 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000670184 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2731075 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2731075 - -
          mouse-6 v1.1 (Illumina) scl52071.1.29_20-S - -
          Illumina Sentrix mouse-6.v1 bead array 670184 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl52071.1.29_20-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 670184 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2884819 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2884819 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline