Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00477177_m1
          See other PSMC3 GE Assays ›
          Gene Symbol:
          Psmc3
          Gene Name:
          proteasome (prosome, macropain) 26S subunit, ATPase 3
          Gene Aliases:
          TBP-1
          Chromosome Location:
          Chr.2: 91054016 - 91059438 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00477177_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Psmc3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008948.2 NP_032974.2 10 - 11 1368 71
          GenBank mRNA AK002542.1 - 10 - 11 1089 71
          AK083043.1 - 1330 71
          AK151370.1 - 10 - 11 1088 71
          AK152018.1 - 10 - 11 1107 71
          AK167002.1 - 10 - 11 1316 71
          AK167061.1 - 10 - 11 1313 71
          AK168263.1 - 9 - 10 1037 71
          AK168616.1 - 10 - 11 1088 71
          BC005783.1 - 10 - 11 1257 3603053 71
          D49686.1 - 10 - 11 1312 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200001366.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Psmc3

          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          blastocyst development
          protein catabolic process
          ER-associated ubiquitin-dependent protein catabolic process
          positive regulation of RNA polymerase II transcriptional preinitiation complex assembly
          positive regulation of proteasomal protein catabolic process
          nucleotide binding
          ATP binding
          hydrolase activity
          TBP-class protein binding
          proteasome-activating ATPase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21420372 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21420368 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21420371 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21420369 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2103429 AK168616.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2103429 BC005783.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2103429 BE989849.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2103429 D49686.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2103429 AK152018.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2121961 AK168616.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2121961 BC005783.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2121961 D49686.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2121961 AK152018.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2121961 AK167002.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 108067 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 779805 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 853514 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 857861 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 128247 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P146060 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2121961 AK168616.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2121961 BC005783.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2121961 D49686.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2121961 AK152018.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2121961 AK167002.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2103429 AK168616.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2103429 BC005783.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2103429 BE989849.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2103429 D49686.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2103429 AK152018.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P280971 AK214668.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P280971 BC005783.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P280971 D49686.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P280971 AK152018.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P280971 AK167002.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420374 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420354 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420389 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420368 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420383 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420376 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420378 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420387 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420357 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420353 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420350 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420371 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420380 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420379 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420361 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420370 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420366 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420351 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420388 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420364 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420385 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420358 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P146060 - -
          Whole Mouse Genome G4122A A_51_P280971 AK214668.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P280971 BC005783.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P280971 D49686.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P280971 AK152018.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P280971 AK167002.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116555 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135086 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P146060 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02144082 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02144078 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21420368 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21420372 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21420369 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21420371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01447967 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02144082 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420350 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02069505 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02144077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420400 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01389774 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01519054 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420349 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01449709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420371 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420402 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420403 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420417 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02029761 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420366 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420411 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02029757 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01578897 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420353 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420379 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420398 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420419 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420374 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420376 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02029768 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02029765 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420405 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420394 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02034540 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01161877 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420393 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420404 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420396 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01389779 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01590528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01550928 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420357 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01519067 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420344 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420421 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01519066 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420380 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420408 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01389789 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02069504 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420383 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10473795 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473796 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473801 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473802 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473798 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473804 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473799 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473797 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473803 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473805 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473806 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473800 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473794 - -
          Affymetrix HT MG-430 PM Array Plate 1416282_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10473793 - -
          Mouse Genome 430 2.0 Array 1416282_PM_at - -
          Mouse Genome 430 2.0 Array 1416282_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19182_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19182_at - -
          Affymetrix Mouse Clariom S Array PSR0200010496.mm.1 NM_008948.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010496.mm.1 NM_008948.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010496.mm.1 TC0200001366.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200010497.mm.1 NM_008948.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010497.mm.1 NM_008948.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010497.mm.1 TC0200001366.mm.1 MTA TC
          Mouse Expression Set 430 1416282_PM_at - -
          Mouse Expression Set 430 1416282_at - -
          Murine Genome U74v2 Set 100225_f_at - -
          Murine Genome U74v2 Set 93734_PM_i_at - -
          Murine Genome U74v2 Set 100225_PM_f_at - -
          Murine Genome U74v2 Set 93734_i_at - -
          Murine Genome U74v2 Set 93735_f_at - -
          Murine Genome U74v2 Set 93735_PM_f_at - -
          Murine Genome U74v2 Set 168979_PM_i_at - -
          Murine Genome U74v2 Set 168979_i_at - -
          Mouse Genome 430A 2.0 Array 1416282_at - -
          Mouse Genome 430A 2.0 Array 1416282_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1416282_at - -
          Affymetrix HT Mouse Genome 430A Array 1416282_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 530679 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2480184 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002480184 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2686571 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2686571 - -
          mouse-6 v1.1 (Illumina) scl019182.11_5-S - -
          Illumina Sentrix mouse-6.v1 bead array 2480184 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl019182.11_5-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2480184 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2686571 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2686571 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl019182.11_5-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline