Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00479148_m1
          See other PRL3C1 GE Assays ›
          Gene Symbol:
          Prl3c1
          Gene Name:
          prolactin family 3, subfamily c, member 1
          Gene Aliases:
          PLP-J, Prlpi, Prlpj, decidualin
          Chromosome Location:
          Chr.13: 27196660 - 27203749 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00479148_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prl3c1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001163218.1 NP_001156690.1 4 - 5 532 69
          NM_013766.2 NP_038794.1 4 - 5 502 69
          GenBank mRNA AB019118.1 - 4 - 5 499 69
          AF150740.1 - 4 - 5 512 69
          AF234637.1 - 4 - 5 502 69
          AK014980.1 - 4 - 5 532 69
          BC120576.1 - 4 - 5 490 40129499 69
          BC120578.1 - 4 - 5 490 40129501 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300000296.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prl3c1

          Panther Classification:

          Molecular Function -

          peptide hormone

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function
          hormone activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966134 AF234637.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966134 BC120576.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966134 BC120578.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966134 NM_013766.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966134 AB019118.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1963046 NM_001163218.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 573231 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 487325 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 449944 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 910674 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 479298 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 73468 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966134 AF234637.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966134 BC120576.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966134 BC120578.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966134 NM_013766.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966134 AB019118.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1963046 NM_001163218.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28901133 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28901114 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28901102 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28901116 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28901104 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28901110 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28901107 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28901125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28901136 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28901129 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28901134 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28901121 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28901108 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28901124 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28901132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28901112 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28901127 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28901113 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28901122 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28901115 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28901128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28901123 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28901118 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28901139 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28901105 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28901111 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28901106 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28901120 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07818169 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07818155 - -
          Whole Mouse Genome G4122A A_52_P281769 AF234637.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P281769 AK014980.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P281769 BC120576.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P281769 BC120578.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P281769 NM_001163218.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109115 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07818169 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10752776 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10752765 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10752755 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10752773 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10752770 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10521845 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10752768 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10521846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28901105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901151 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901165 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901177 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901170 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11061565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901108 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901140 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901114 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901111 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901134 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901162 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901173 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10869223 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901128 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901125 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901176 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10666984 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901127 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901133 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901178 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901112 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901143 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901172 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901115 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901158 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901118 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901123 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901146 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901166 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901122 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901110 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901148 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901120 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901113 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901164 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901153 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901109 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28901154 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10404289 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404291 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404293 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404292 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404288 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404290 - -
          Affymetrix HT MG-430 PM Array Plate 1449830_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10404287 - -
          Mouse Genome 430 2.0 Array 1449830_PM_at - -
          Mouse Genome 430 2.0 Array 1449830_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 27372_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 27372_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1300001839.mm.1 NM_001163218.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001839.mm.1 NM_001163218.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001839.mm.1 NM_013766.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001839.mm.1 NM_013766.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001839.mm.1 TC1300000296.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300001841.mm.1 NM_001163218.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001841.mm.1 NM_001163218.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001841.mm.1 NM_013766.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001841.mm.1 NM_013766.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001841.mm.1 TC1300000296.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300001840.mm.1 NM_001163218.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001840.mm.1 NM_001163218.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001840.mm.1 NM_013766.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001840.mm.1 NM_013766.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001840.mm.1 TC1300000296.mm.1 MTA TC
          Mouse Expression Set 430 1449830_PM_at - -
          Mouse Expression Set 430 1449830_at - -
          Murine Genome U74v2 Set 92274_at - -
          Murine Genome U74v2 Set 92274_PM_at - -
          Mouse Genome 430A 2.0 Array 1449830_at - -
          Mouse Genome 430A 2.0 Array 1449830_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449830_at - -
          Affymetrix HT Mouse Genome 430A Array 1449830_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 699540 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2850110 - -
          Illumina mouse-6 v1.0 expression beadchip 2850110 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002850110 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2724496 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2724496 - -
          mouse-6 v1.1 (Illumina) scl44967.6.1_184-S - -
          Illumina Sentrix mouse-6.v1 bead array 2850110 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl44967.6.1_184-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2850110 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2724496 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2724496 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline