Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00480922_m1
          See other ZNRD1 GE Assays ›
          Gene Symbol:
          Znrd1
          Gene Name:
          zinc ribbon domain containing, 1
          Gene Aliases:
          1110014N07Rik, Rpa12
          Chromosome Location:
          Chr.17: 36954349 - 36958520 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00480922_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Znrd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023162.4 NP_075651.1 3 - 4 495 111
          GenBank mRNA AF230339.1 - 3 - 4 368 111
          AK003714.1 - 3 - 4 496 111
          AK019152.1 - 4 - 5 472 111
          BC043016.1 - 4 - 5 432 5683859 111

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700002042.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Znrd1

          Panther Classification:

          Molecular Function -

          DNA-directed RNA polymerase RNA metabolism protein nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          termination of RNA polymerase I transcription
          RNA polymerase I activity
          nucleic acid binding
          DNA-directed RNA polymerase activity
          zinc ion binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31166480 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286142 NM_023162.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286142 AF230339.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286142 AK003714.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286142 AK019152.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286142 BC043016.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286142 NM_023162.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286142 AF230339.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286142 AK003714.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286142 AK019152.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286142 BC043016.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 285482 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 4381 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 762120 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P122123 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286142 NM_023162.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286142 AF230339.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286142 AK003714.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286142 AK019152.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286142 BC043016.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286142 NM_023162.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286142 AF230339.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286142 AK003714.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286142 AK019152.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286142 BC043016.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P203758 NM_023162.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P203758 AF230339.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P203758 AK003714.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P203758 AK005976.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P203758 AK019152.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P286142 NM_023162.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P286142 AF230339.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P286142 AK003714.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P286142 AK019152.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P286142 BC043016.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P286142 NM_023162.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P286142 AF230339.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P286142 AK003714.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P286142 AK019152.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P286142 BC043016.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166470 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166466 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166460 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166473 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166469 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166462 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166458 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166478 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166459 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166475 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166480 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166465 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P122123 - -
          Whole Mouse Genome G4122A A_52_P203758 NM_023162.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P203758 AF230339.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P203758 AK003714.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P203758 AK005976.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P203758 AK019152.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286142 NM_023162.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286142 AF230339.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286142 AK003714.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286142 AK019152.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286142 BC043016.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286142 NM_023162.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286142 AF230339.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286142 AK003714.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286142 AK019152.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286142 BC043016.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138307 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109783 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P122123 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13149274 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13149276 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13149264 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13149269 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13149277 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13149267 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31166480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166480 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166458 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166462 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166471 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166459 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166465 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13540192 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13317221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166470 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166467 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166480 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13149267 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166460 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166475 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166463 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166473 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10450822 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450820 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450823 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450821 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450825 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450827 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450826 - -
          Affymetrix HT MG-430 PM Array Plate 1422517_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10450819 - -
          Mouse Genome 430 2.0 Array 1422517_a_at - -
          Mouse Genome 430 2.0 Array 1422517_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66136_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66136_at - -
          Affymetrix Mouse Clariom S Array PSR1700019458.mm.1 NM_023162.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019458.mm.1 NM_023162.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019458.mm.1 TC1700002042.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019460.mm.1 NM_023162.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019460.mm.1 NM_023162.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019460.mm.1 TC1700002042.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019459.mm.1 NM_023162.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019459.mm.1 NM_023162.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019459.mm.1 TC1700002042.mm.1 MTA TC
          Mouse Expression Set 430 1422517_a_at - -
          Mouse Expression Set 430 1422517_PM_a_at - -
          Murine Genome U74v2 Set 96711_PM_at - -
          Murine Genome U74v2 Set 96711_at - -
          Murine Genome U74v2 Set 167740_i_at - -
          Murine Genome U74v2 Set 167740_PM_i_at - -
          Mouse Genome 430A 2.0 Array 1422517_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1422517_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1422517_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1422517_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 927485 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5340739 - -
          Illumina mouse-6 v1.0 expression beadchip 4150373 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004150373 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005340739 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1235476 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1247048 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1235476 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1247048 - -
          mouse-6 v1.1 (Illumina) scl49941.4.1_39-S - -
          mouse-6 v1.1 (Illumina) scl0001598.1_20-S - -
          Illumina Sentrix mouse-6.v1 bead array 4150373 - -
          Illumina Sentrix mouse-6.v1 bead array 5340739 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0001598.1_20-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl49941.4.1_39-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4150373 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5340739 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1247048 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2971486 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2971481 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1247048 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2971486 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2971481 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl49941.4.1_39-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline