Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00481276_m1
          See other TMEM14C GE Assays ›
          Gene Symbol:
          Tmem14c
          Gene Name:
          transmembrane protein 14C
          Gene Aliases:
          1110021D01Rik, AU020193, HSPC194
          Chromosome Location:
          Chr.13: 41016250 - 41022582 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00481276_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem14c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025387.3 NP_079663.1 3 - 4 287 113
          GenBank mRNA AK003891.1 - 3 - 4 236 113
          AK008495.1 - 2 - 3 177 113
          AK008878.1 - 3 - 4 209 113
          AK010609.1 - 3 - 4 247 113
          AK012101.1 - 3 - 4 234 113
          AK019395.1 - 3 - 4 236 113
          AK050688.1 - 3 - 4 222 113
          AK089021.1 - 2 - 3 176 113
          BC025854.1 - 3 - 4 188 5250659 113
          BC055862.1 - 3 - 4 211 3492090 113
          BC069899.1 - 2 - 3 933 113

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300000463.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem14c

          Gene Ontology Categories:

          Function(s) Process(es)

          heme biosynthetic process
          mitochondrial transport
          erythrocyte differentiation
          regulation of heme biosynthetic process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28975562 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28975560 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28975561 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28975559 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P338600 BC025854.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P338600 BC055862.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P338600 BC069899.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P338600 CO039120.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P338600 AK003891.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P338600 BC025854.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P338600 BC055862.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P338600 BC069899.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P338600 CO039120.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P338600 AK003891.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 308417 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 352251 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 648003 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 926980 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 35612 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P116858 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P338600 BC025854.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P338600 BC055862.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P338600 BC069899.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P338600 CO039120.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P338600 AK003891.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P338600 BC025854.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P338600 BC055862.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P338600 BC069899.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P338600 CO039120.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P338600 AK003891.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28975548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28975571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28975558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28975575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28975579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28975577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28975578 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28975559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28975566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28975567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28975549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28975574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28975564 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28975569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28975557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28975545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28975560 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28975552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28975562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28975555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28975551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28975565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28975553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28975546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28975576 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28975550 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02673651 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P116858 - -
          Whole Mouse Genome G4122A A_51_P338600 BC025854.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P338600 BC055862.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P338600 BC069899.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P338600 CO039120.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P338600 AK003891.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P338600 BC025854.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P338600 BC055862.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P338600 BC069899.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P338600 CO039120.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P338600 AK003891.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106439 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120083 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P116858 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11000889 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11000886 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11025662 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11025669 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11025658 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11025656 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11025667 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11000890 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28975560 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28975561 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28975562 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28975559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28975575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11097140 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11025659 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11025663 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28975598 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28975615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10937953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11099835 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11099828 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28975613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28975607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28975569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11025656 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28975560 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28975610 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28975549 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10893812 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28975553 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28975596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10677863 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10474261 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11000886 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28975555 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28975567 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28975577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28975595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11099846 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28975574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28975591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28975575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28975547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10656408 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11099842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11099832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28975597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28975589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28975606 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28975604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28975565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28975571 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28975608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10792805 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28975603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28975588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28975586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28975548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11099837 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28975600 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28975557 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28975558 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10404733 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404734 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404736 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404735 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404732 - -
          Affymetrix HT MG-430 PM Array Plate 1416479_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10404731 - -
          Mouse Genome 430 2.0 Array 1416479_a_at - -
          Mouse Genome 430 2.0 Array 1416479_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66154_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66154_at - -
          Affymetrix Mouse Clariom S Array PSR1300002749.mm.1 NM_025387.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002749.mm.1 NM_025387.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002749.mm.1 TC1300000463.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300002751.mm.1 NM_025387.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002751.mm.1 NM_025387.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002751.mm.1 TC1300000463.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300002752.mm.1 NM_025387.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002752.mm.1 NM_025387.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002752.mm.1 TC1300000463.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300002750.mm.1 NM_025387.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002750.mm.1 NM_025387.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002750.mm.1 TC1300000463.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300002747.mm.1 NM_025387.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002747.mm.1 NM_025387.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002747.mm.1 TC1300000463.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300002748.mm.1 NM_025387.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002748.mm.1 NM_025387.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002748.mm.1 TC1300000463.mm.1 MTA TC
          Mouse Expression Set 430 1416479_a_at - -
          Mouse Expression Set 430 1416479_PM_a_at - -
          Murine Genome U74v2 Set 96353_PM_at - -
          Murine Genome U74v2 Set 96353_at - -
          Mouse Genome 430A 2.0 Array 1416479_a_at - -
          Mouse Genome 430A 2.0 Array 1416479_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1416479_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1416479_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 916116 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 670647 - -
          MouseWG-6 v2 Expression BeadChip 3940193 - -
          Illumina mouse-6 v1.0 expression beadchip 3940193 - -
          Illumina mouse-6 v1.0 expression beadchip 670647 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000670647 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003940193 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2435206 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2472110 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2435206 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2472110 - -
          mouse-6 v1.1 (Illumina) scl0003742.1_4-S - -
          mouse-6 v1.1 (Illumina) scl066154.6_169-S - -
          Illumina Sentrix mouse-6.v1 bead array 670647 - -
          Illumina Sentrix mouse-6.v1 bead array 3940193 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0003742.1_4-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl066154.6_169-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 670647 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3940193 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2472110 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2849680 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2435206 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2849680 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2472110 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2435206 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0003742.1_4-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl066154.6_169-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline