Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00482543_m1
          See other TSPAN31 GE Assays ›
          Gene Symbol:
          Tspan31
          Gene Name:
          tetraspanin 31
          Gene Aliases:
          2700085A14Rik, AW742554, Sas
          Chromosome Location:
          Chr.10: 127067290 - 127070261 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00482543_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tspan31
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025982.4 NP_080258.1 1 - 2 146 58
          GenBank mRNA AK012457.1 - 1 - 2 147 58
          AK013523.1 - 1 - 2 159 58
          AK014183.1 - 1 - 2 145 58
          AK075962.1 - 1 - 2 147 58
          BC023306.1 - 1 - 2 123 4165774 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000003096.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tspan31

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407074 AK012457.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407074 AK012457.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 694392 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 814468 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 970012 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P134538 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407074 AK012457.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407074 AK012457.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533888 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533904 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533911 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533906 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533875 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533902 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533903 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533882 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533907 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533868 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533859 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533889 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533910 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533897 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533890 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533866 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533896 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533894 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533887 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533891 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533892 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533898 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27533851 - -
          Whole Mouse Genome G4122A A_51_P407074 AK012457.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407074 AK012457.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133053 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08323692 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08437198 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08437200 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08437201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27533894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533876 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08323692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533879 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533868 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08903907 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08380748 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08606902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08501998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533852 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08750467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08437200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533872 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27533851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08323680 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10373028 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373029 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373035 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373034 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373033 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373031 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373032 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373030 - -
          Affymetrix HT MG-430 PM Array Plate 1455853_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1430029_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1416556_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10373027 - -
          Mouse Genome 430 2.0 Array 1455853_x_at - -
          Mouse Genome 430 2.0 Array 1416556_at - -
          Mouse Genome 430 2.0 Array 1430029_a_at - -
          Mouse Genome 430 2.0 Array 1455853_PM_x_at - -
          Mouse Genome 430 2.0 Array 1430029_PM_a_at - -
          Mouse Genome 430 2.0 Array 1416556_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67125_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67125_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1000023048.mm.1 NM_025982.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023048.mm.1 NM_025982.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023048.mm.1 TC1000003096.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000023049.mm.1 NM_025982.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023049.mm.1 NM_025982.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023049.mm.1 TC1000003096.mm.1 MTA TC
          Mouse Expression Set 430 1455853_PM_x_at - -
          Mouse Expression Set 430 1416556_PM_at - -
          Mouse Expression Set 430 1430029_a_at - -
          Mouse Expression Set 430 1416556_at - -
          Mouse Expression Set 430 1455853_x_at - -
          Mouse Expression Set 430 1430029_PM_a_at - -
          Murine Genome U74v2 Set 98144_PM_f_at - -
          Murine Genome U74v2 Set 98144_f_at - -
          Murine Genome U74v2 Set 170314_PM_f_at - -
          Murine Genome U74v2 Set 170314_f_at - -
          Mouse Genome 430A 2.0 Array 1455853_x_at - -
          Mouse Genome 430A 2.0 Array 1416556_PM_at - -
          Mouse Genome 430A 2.0 Array 1430029_a_at - -
          Mouse Genome 430A 2.0 Array 1416556_at - -
          Mouse Genome 430A 2.0 Array 1430029_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1455853_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1430029_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1430029_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1416556_at - -
          Affymetrix HT Mouse Genome 430A Array 1416556_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1455853_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1455853_x_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2030333 - -
          Illumina mouse-6 v1.0 expression beadchip 6520279 - -
          Illumina mouse-6 v1.0 expression beadchip 2030333 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002030333 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006520279 - -
          mouse-6 v1.1 (Illumina) scl37402.3_73-S - -
          Illumina Sentrix mouse-6.v1 bead array 2030333 - -
          Illumina Sentrix mouse-6.v1 bead array 6520279 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37402.3_73-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0067125.2_158-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2030333 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6520279 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2757599 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2964076 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2964076 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2757599 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl37402.3_73-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline