Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00482759_m1
          See other PAXBP1 GE Assays ›
          Gene Symbol:
          Paxbp1
          Gene Name:
          PAX3 and PAX7 binding protein 1
          Gene Aliases:
          1810007M14Rik, C21orf66, Gcfc, Gcfc1, Pax3/7bp
          Chromosome Location:
          Chr.16: 91014037 - 91044734 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00482759_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Paxbp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026110.2 NP_080386.3 7 - 8 1414 102
          GenBank mRNA AK007365.1 - 1 - 2 102 102
          AY033907.1 - 7 - 8 1199 102
          AY033908.1 - 7 - 8 1199 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600002061.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK007365.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Paxbp1

          Panther Classification:

          Molecular Function -

          chromatin/chromatin-binding, or -regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          muscle organ development
          regulation of skeletal muscle satellite cell proliferation
          positive regulation of histone methylation
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of myoblast proliferation
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          transcription factor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P100232 AY033908.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P100232 BC014838.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P100232 BC027145.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P100232 AY033907.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P100232 AK011477.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P478745 AK029496.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P478745 NM_026110.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P478745 AK029496.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P478745 NM_026110.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 670690 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 445036 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 645360 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 33726 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 27217 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 681516 AK082860.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 516410 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 226287 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 800167 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 972877 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 257627 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 307658 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 847410 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 693288 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 198485 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 485398 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 27704 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 669473 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 61959 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 156074 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 120628 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106731 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P171058 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P100232 AY033908.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P100232 BC014838.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P100232 BC027145.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P100232 AY033907.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P100232 AK011477.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P478745 AK029496.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P478745 NM_026110.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P478745 AK029496.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P478745 NM_026110.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08681881 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08681893 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106731 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P171058 - -
          Whole Mouse Genome G4122A A_52_P478745 AK029496.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P478745 NM_026110.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P478745 AK029496.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P478745 NM_026110.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P317166 AK007365.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100232 AY033908.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100232 BC014838.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100232 BC027145.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100232 AY033907.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100232 AK011477.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08681893 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106731 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P171058 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960658 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960528 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12963889 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960488 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12621463 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960474 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12647718 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960553 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960509 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12532182 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960637 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12611198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960463 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12614549 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12712529 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960600 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960522 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960535 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12963893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960566 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960521 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960583 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960610 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960524 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12736050 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960496 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12906365 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960465 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12499294 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960617 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960552 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960598 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960470 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960629 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960489 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12611200 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12587784 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960505 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960557 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960471 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960501 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960627 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960568 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960481 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960656 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960517 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960498 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960660 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960650 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960559 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12564126 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12653012 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960643 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960562 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960537 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960468 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960479 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960483 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960525 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960466 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960473 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960570 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960493 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960551 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960495 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960508 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960526 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960542 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960512 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960654 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960646 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12879749 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960539 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12611201 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960534 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30960581 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10440897 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440888 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440885 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440898 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440900 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440883 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440887 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440886 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440899 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440889 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440882 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440895 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440893 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440894 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440901 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440884 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440891 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440892 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440902 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440896 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440890 - -
          Affymetrix HT MG-430 PM Array Plate 1418008_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1418009_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1418007_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10440881 - -
          Mouse Genome 430 2.0 Array 1418009_at - -
          Mouse Genome 430 2.0 Array 1418009_PM_at - -
          Mouse Genome 430 2.0 Array 1418007_at - -
          Mouse Genome 430 2.0 Array 1418007_PM_at - -
          Mouse Genome 430 2.0 Array 1418008_at - -
          Mouse Genome 430 2.0 Array 1418008_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67367_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67367_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1600015871.mm.1 NM_026110.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600015871.mm.1 NM_026110.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600015871.mm.1 TC1600002061.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600015872.mm.1 NM_026110.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600015872.mm.1 NM_026110.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600015872.mm.1 TC1600002061.mm.1 MTA TC
          Mouse Expression Set 430 1418007_at - -
          Mouse Expression Set 430 1418008_PM_at - -
          Mouse Expression Set 430 1418009_PM_at - -
          Mouse Expression Set 430 1418008_at - -
          Mouse Expression Set 430 1418009_at - -
          Mouse Expression Set 430 1418007_PM_at - -
          Murine Genome U74v2 Set 92233_at - -
          Murine Genome U74v2 Set 92233_PM_at - -
          Mouse Genome 430A 2.0 Array 1418007_at - -
          Mouse Genome 430A 2.0 Array 1418008_at - -
          Mouse Genome 430A 2.0 Array 1418008_PM_at - -
          Mouse Genome 430A 2.0 Array 1418009_PM_at - -
          Mouse Genome 430A 2.0 Array 1418007_PM_at - -
          Mouse Genome 430A 2.0 Array 1418009_at - -
          Affymetrix HT Mouse Genome 430A Array 1418008_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418007_at - -
          Affymetrix HT Mouse Genome 430A Array 1418009_at - -
          Affymetrix HT Mouse Genome 430A Array 1418008_at - -
          Affymetrix HT Mouse Genome 430A Array 1418007_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418009_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 774601 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 610528 - -
          Illumina mouse-6 v1.0 expression beadchip 610528 - -
          Illumina mouse-6 v1.0 expression beadchip 1770504 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000610528 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001770504 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2681311 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1228596 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2681311 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1228596 - -
          mouse-6 v1.1 (Illumina) scl0001835.1_46-S - -
          mouse-6 v1.1 (Illumina) scl48212.18.1_6-S - -
          Illumina Sentrix mouse-6.v1 bead array 610528 - -
          Illumina Sentrix mouse-6.v1 bead array 1770504 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0001835.1_46-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl48212.18.1_6-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1770504 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 610528 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2681311 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1228596 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2681311 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1228596 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline