Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00485539_m1
          See other RAPSN GE Assays ›
          Gene Symbol:
          Rapsn
          Gene Name:
          receptor-associated protein of the synapse
          Gene Aliases:
          43kDa, Nraps, Raps, rapsyn
          Chromosome Location:
          Chr.2: 91035466 - 91045729 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00485539_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rapsn
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009023.3 NP_033049.2 2 - 3 698 67
          GenBank mRNA BC111863.1 - 2 - 3 654 40046186 67
          J03962.1 - 2 - 3 634 67
          X15788.1 - 2 - 3 687 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200001365.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rapsn

          Gene Ontology Categories:

          Function(s) Process(es)

          chemical synaptic transmission
          synaptic transmission, cholinergic
          positive regulation of neuron apoptotic process
          skeletal muscle acetylcholine-gated channel clustering
          positive regulation of neuromuscular synaptic transmission
          regulation of postsynaptic membrane organization
          zinc ion binding
          acetylcholine receptor binding
          ionotropic glutamate receptor binding
          protein anchor
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2087322 J03962.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2087322 X15788.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2087322 NM_009023.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 863970 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 662636 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 697847 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 116801 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 830754 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 823166 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 25108 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 132175 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131454 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140844 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2087322 J03962.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2087322 X15788.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2087322 NM_009023.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P385974 BC111863.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P385974 J03962.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P385974 X15788.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P385974 NM_009023.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P385974 AK009672.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420270 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420289 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420258 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420284 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420291 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420260 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420266 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420277 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420285 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420286 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420262 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420265 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420278 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420293 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420281 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420288 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420275 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420257 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420249 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21420282 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140844 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131454 - -
          Whole Mouse Genome G4122A A_51_P385974 BC111863.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P385974 J03962.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P385974 X15788.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P385974 NM_009023.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P385974 AK009672.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119052 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125066 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131454 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140844 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02068475 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01294841 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01294845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21420261 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420310 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01900033 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01900031 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01618330 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420302 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420267 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01294840 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01618327 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420266 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01950264 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01955554 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01953762 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420299 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01388867 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02147901 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01558292 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01955555 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01294849 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01294845 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420301 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01294843 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02230959 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01294835 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420252 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01294852 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02132420 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420317 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01444351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420316 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02147904 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420276 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420315 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01444354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01240828 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420270 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01444358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01294837 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420285 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02054445 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420275 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420281 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420271 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420257 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420291 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21420311 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10473784 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473786 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473792 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473791 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473785 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473788 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473789 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473790 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473787 - -
          Affymetrix HT MG-430 PM Array Plate 1449331_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10473783 - -
          Mouse Genome 430 2.0 Array 1457151_PM_at - -
          Mouse Genome 430 2.0 Array 1449331_PM_a_at - -
          Mouse Genome 430 2.0 Array 1449331_a_at - -
          Mouse Genome 430 2.0 Array 1457151_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19400_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19400_PM_at - -
          Affymetrix Murine Genome 6500A Array Msa11260_at - -
          Affymetrix Mouse Clariom S Array PSR0200010459.mm.1 NM_009023.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010459.mm.1 NM_009023.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010459.mm.1 TC0200001365.mm.1 MTA TC
          Mouse Expression Set 430 1449331_PM_a_at - -
          Mouse Expression Set 430 1449331_a_at - -
          Murine Genome U74v2 Set 102300_PM_at - -
          Murine Genome U74v2 Set 102300_at - -
          Mouse Genome 430A 2.0 Array 1449331_a_at - -
          Mouse Genome 430A 2.0 Array 1449331_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1449331_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1449331_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 694459 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 610110 - -
          Illumina mouse-6 v1.0 expression beadchip 2370162 - -
          Illumina mouse-6 v1.0 expression beadchip 610110 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000610110 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002370162 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1235236 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2723095 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1235236 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2723095 - -
          mouse-6 v1.1 (Illumina) scl0003276.1_11-S - -
          mouse-6 v1.1 (Illumina) scl20648.8.1_43-S - -
          Illumina Sentrix mouse-6.v1 bead array 2370162 - -
          Illumina Sentrix mouse-6.v1 bead array 610110 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0003276.1_11-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl20648.8.1_43-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 610110 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2370162 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2967654 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2723095 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1235236 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1235236 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2967654 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2723095 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline