Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00487416_m1
          See other FKBP7 GE Assays ›
          Gene Symbol:
          Fkbp7
          Gene Name:
          FK506 binding protein 7
          Gene Aliases:
          23kDa, FKBP-7, FKBP23
          Chromosome Location:
          Chr.2: 76663034 - 76673139 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00487416_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fkbp7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010222.2 NP_034352.1 1 - 2 298 78
          GenBank mRNA AF040252.1 - 1 - 2 270 78
          AK042208.1 - 1 - 2 286 78
          AK138924.1 - 1 - 2 214 78
          BC032961.1 - 1 - 2 255 1347695 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003806.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fkbp7

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          protein peptidyl-prolyl isomerization
          protein folding
          chaperone-mediated protein folding
          peptidyl-prolyl cis-trans isomerase activity
          calcium ion binding
          protein binding
          FK506 binding
          isomerase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21351694 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21351693 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21351695 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P415607 AF040252.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P415607 AF040252.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 599769 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 790402 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 307136 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 57801 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 184259 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 772266 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P129642 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P173089 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P415607 AF040252.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P415607 AF040252.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P415607 AF040252.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P415607 AF040252.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21351680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21351681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21351674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21351677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21351675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21351669 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21351676 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21351691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21351673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21351686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21351695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21351683 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21351678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21351689 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21351667 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21351671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21351692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21351682 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21351696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21351687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21351679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21351685 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21351668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21351684 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00409814 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P173089 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P129642 - -
          Whole Mouse Genome G4122A A_51_P415607 AF040252.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P415607 AF040252.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P129642 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P173089 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01703951 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01703932 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01703922 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01703934 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01703944 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01703925 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01703924 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01692302 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21351695 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21351694 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21351693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21351711 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21351677 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21351669 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21351681 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21351652 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21351651 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21351659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21351665 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21351689 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21351657 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01703944 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21351642 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01645493 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01478753 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21351684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21351675 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21351653 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01703951 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01703920 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01645495 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21351620 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21351671 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01750919 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21351676 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21351674 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21351668 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21351679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21351645 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21351687 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21351644 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21351667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21351647 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02045710 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21351672 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21351658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01703949 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21351655 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02045712 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10483869 - -
          Affymetrix Mouse Gene 1.0 ST Array 10483867 - -
          Affymetrix Mouse Gene 1.0 ST Array 10483870 - -
          Affymetrix Mouse Gene 1.0 ST Array 10483866 - -
          Affymetrix HT MG-430 PM Array Plate 1446078_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1416803_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10483865 - -
          Mouse Genome 430 2.0 Array 1416803_at - -
          Mouse Genome 430 2.0 Array 1416803_PM_at - -
          Mouse Genome 430 2.0 Array 1446078_at - -
          Mouse Genome 430 2.0 Array 1446078_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14231_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14231_at - -
          Affymetrix Mouse Clariom S Array PSR0200032508.mm.1 NM_010222.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200032508.mm.1 NM_010222.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200032508.mm.1 TC0200003806.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200032507.mm.1 NM_010222.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200032507.mm.1 NM_010222.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200032507.mm.1 TC0200003806.mm.1 MTA TC
          Mouse Expression Set 430 1416803_at - -
          Mouse Expression Set 430 1416803_PM_at - -
          Mouse Expression Set 430 1446078_at - -
          Mouse Expression Set 430 1446078_PM_at - -
          Murine Genome U74v2 Set 104746_at - -
          Murine Genome U74v2 Set 104746_PM_at - -
          Mouse Genome 430A 2.0 Array 1416803_at - -
          Mouse Genome 430A 2.0 Array 1416803_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1416803_at - -
          Affymetrix HT Mouse Genome 430A Array 1416803_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1446078_at - -
          Affymetrix HT Mouse Genome 430B Array 1446078_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 618976 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100520195 - -
          MouseWG-6 v2 Expression BeadChip 6020064 - -
          Illumina mouse-6 v1.0 expression beadchip 101690315 - -
          Illumina mouse-6 v1.0 expression beadchip 6020064 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006020064 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1227875 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1224152 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1227875 - -
          mouse-6 v1.1 (Illumina) scl19106.4.1_16-S - -
          Illumina Sentrix mouse-6.v1 bead array 6020064 - -
          Illumina Sentrix mouse-6.v1 bead array 101690315 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl19106.4.1_16-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101690315 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6020064 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1227875 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1224152 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1227875 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline