Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00488750_m1
          See other CEACAM9 GE Assays ›
          Gene Symbol:
          Ceacam9
          Gene Name:
          carcinoembryonic antigen-related cell adhesion molecule 9
          Gene Aliases:
          AA410097, AW545709, Cea-5, Cea5, mmCGM8
          Chromosome Location:
          Chr.7: 16721929 - 16726110 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00488750_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ceacam9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011927.4 NP_036057.1 2 - 3 461 74
          GenBank mRNA AK049543.1 - 2 - 3 433 74
          L49180.1 - 2 - 3 461 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000233.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ceacam9

          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P523897 NM_011927.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P523897 AK049543.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P523897 L49180.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P523897 NM_011927.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P523897 AK049543.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P523897 L49180.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 590557 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 400592 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 11537 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139096 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P523897 NM_011927.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P523897 AK049543.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P523897 L49180.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P523897 NM_011927.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P523897 AK049543.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P523897 L49180.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24982932 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24982934 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24982976 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24982982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24982952 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24982980 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24982986 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24982978 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24982941 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24982955 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24982962 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24982977 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24982944 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24982954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24982961 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24982983 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24982956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24982963 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24982933 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139096 - -
          Whole Mouse Genome G4122A A_52_P523897 NM_011927.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P523897 AK049543.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P523897 L49180.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P523897 NM_011927.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P523897 AK049543.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P523897 L49180.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112796 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139096 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06657041 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05952964 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06087370 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06087369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24982988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24982941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24982982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24982974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24982911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24982986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24982930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24982952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24982913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24982994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24982962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24982932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24982995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24982929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24982910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24982992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24982980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24982914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24982961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24982917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24982998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24982976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24982925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24982927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24982953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24982928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24982987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24982978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24982977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24982989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24982956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24982999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24982908 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24982944 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24982963 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06087369 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05966760 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24982995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24982919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24982925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06614575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24982941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05952964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06084183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06084185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06608558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06614578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24982974 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24982980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06614574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24982952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06614582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24982934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06087370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24982954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05963840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24982988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06084182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24982998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06614588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24982982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24982940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05963841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06084186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24982953 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24982923 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05963843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06438157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06614580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24982999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05995752 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10550324 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550325 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550323 - -
          Affymetrix HT MG-430 PM Array Plate 1418218_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10550322 - -
          Mouse Genome 430 2.0 Array 1418218_at - -
          Mouse Genome 430 2.0 Array 1418218_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 26368_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 26368_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700001693.mm.1 TC0700000233.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700001693.mm.1 NM_011927.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001693.mm.1 NM_011927.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001692.mm.1 TC0700000233.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700001692.mm.1 NM_011927.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001692.mm.1 NM_011927.4 Refseq
          Mouse Expression Set 430 1418218_at - -
          Mouse Expression Set 430 1418218_PM_at - -
          Murine Genome U74v2 Set 104159_at - -
          Murine Genome U74v2 Set 104159_PM_at - -
          Mouse Genome 430A 2.0 Array 1418218_at - -
          Mouse Genome 430A 2.0 Array 1418218_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418218_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418218_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 324349 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 580551 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000580551 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2703161 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2703161 - -
          mouse-6 v1.1 (Illumina) scl33049.3.1_29-S - -
          Illumina Sentrix mouse-6.v1 bead array 580551 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl33049.3.1_29-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 580551 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2703161 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2703161 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline