Search
Search

Sfmbt1| Gene Symbol: Sfmbt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166531.1 | NP_001160003.1 | 10 - 11 | 1321 | 80 | |
| NM_001166532.1 | NP_001160004.1 | 9 - 10 | 1415 | 80 | ||
| NM_019460.2 | NP_062333.1 | 9 - 10 | 1272 | 80 | ||
| GenBank mRNA | AB032165.1 | - | 8 - 9 | 1117 | 80 | |
| AK142330.1 | - | 9 - 10 | 1299 | 80 | ||
| AK154826.1 | - | 9 - 10 | 1417 | 80 | ||
| AK160573.1 | - | 10 - 11 | 1321 | 80 | ||
| BC018217.1 | - | 9 - 10 | 1283 | 80 | ||
| BC036972.1 | - | 9 - 10 | 1342 | 80 | ||
| BC066188.1 | - | 9 - 10 | 1225 | 30476111 | 80 | |
Set Membership: |
|