Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00491159_m1
          See other RNF25 GE Assays ›
          Gene Symbol:
          Rnf25
          Gene Name:
          ring finger protein 25
          Gene Aliases:
          0610009H16Rik, AO7
          Chromosome Location:
          Chr.1: 74593748 - 74601431 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00491159_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnf25
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AF171060.1 - 5 - 6 361 101
          AK002399.1 - 5 - 6 382 101
          AK011171.1 - 2 - 3 90 101
          AK019236.1 - 2 - 3 88 101
          AK171752.1 - 5 - 6 359 101
          BC022715.1 - 5 - 6 347 101
          BC037100.1 - 5 - 6 372 101
          BC048821.1 - 5 - 6 369 3966642 101
          BG081026.2 - 5 - 6 328 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100002588.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20349522 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20349520 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20349521 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2122898 BC022715.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2122898 BC037100.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2122898 BC048821.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2122898 AK171752.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2122898 NM_021313.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 591273 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 308435 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 911788 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 600389 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 36486 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 197771 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 18657 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160121 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2122898 BC022715.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2122898 BC037100.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2122898 BC048821.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2122898 AK171752.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2122898 NM_021313.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P249611 BC022715.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P249611 BC037100.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P249611 BC048821.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P249611 AK171752.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P249611 NM_021313.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349490 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349508 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349522 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349494 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349510 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04148287 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P160121 - -
          Whole Mouse Genome G4122A A_52_P249611 BC022715.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P249611 BC037100.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P249611 BC048821.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P249611 AK171752.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P249611 NM_021313.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108946 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108701 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P160121 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00951718 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00951719 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00951713 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00951709 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20349522 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20349521 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20349520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349478 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20349507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20349496 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20349480 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01104019 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20349484 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20349470 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20349508 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20349491 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20349495 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01104022 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00951710 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20349482 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20349473 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20349520 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20349478 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20349510 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20349494 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20349486 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20349503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20349489 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20349472 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20349519 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20349475 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20349488 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00884527 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20349518 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00470211 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00999496 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20349497 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20349516 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00920405 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20349511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00951718 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20349501 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20349493 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20349498 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10355629 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355636 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355630 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355637 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355634 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355631 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355638 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355635 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355632 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355633 - -
          Affymetrix HT MG-430 PM Array Plate 1448674_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10355628 - -
          Mouse Genome 430 2.0 Array 1442108_PM_at - -
          Mouse Genome 430 2.0 Array 1442108_at - -
          Mouse Genome 430 2.0 Array 1448674_PM_at - -
          Mouse Genome 430 2.0 Array 1448674_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 57751_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 57751_at - -
          Affymetrix Mouse Clariom S Array PSR0100020912.mm.1 NM_021313.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100020912.mm.1 TC0100002588.mm.1 MTA TC
          Mouse Expression Set 430 1448674_PM_at - -
          Mouse Expression Set 430 1448674_at - -
          Murine Genome U74v2 Set 165634_PM_at - -
          Murine Genome U74v2 Set 165634_at - -
          Mouse Genome 430A 2.0 Array 1448674_at - -
          Mouse Genome 430A 2.0 Array 1448674_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448674_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448674_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 703804 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6760563 - -
          MouseWG-6 v2 Expression BeadChip 2120632 - -
          Illumina mouse-6 v1.0 expression beadchip 6760563 - -
          Illumina mouse-6 v1.0 expression beadchip 2120632 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006760563 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002120632 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2607369 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2736369 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2736369 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2607369 - -
          mouse-6 v1.1 (Illumina) scl000751.1_62-S - -
          mouse-6 v1.1 (Illumina) scl16611.11.1_62-S - -
          Illumina Sentrix mouse-6.v1 bead array 6760563 - -
          Illumina Sentrix mouse-6.v1 bead array 2120632 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl16611.11.1_62-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl000751.1_62-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6760563 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2120632 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2863905 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2607369 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2863903 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2607369 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2863905 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2863903 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl000751.1_62-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline