Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00493258_s1
          See other OLFR16 GE Assays ›
          Gene Symbol:
          Olfr16
          Gene Name:
          olfactory receptor 16
          Gene Aliases:
          MOR23, MOR26713
          Chromosome Location:
          Chr.1: 172956768 - 172957817 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00493258_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr16
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008763.2 NP_032789.1 1 480 82
          GenBank mRNA BC119298.1 - 1 480 8734047 82
          BC119300.1 - 1 480 8734049 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100001625.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr16

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2119184 BC119298.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2119184 BC119300.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2119184 NM_008763.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P177213 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2119184 BC119298.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2119184 BC119300.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2119184 NM_008763.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P416927 BC119298.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P416927 BC119300.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P416927 NM_008763.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835708 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835707 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835710 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835731 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835706 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835712 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835688 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835715 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835727 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835725 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835733 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835720 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835729 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835700 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835716 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835718 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835735 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835719 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835714 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835698 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835711 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835736 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835732 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835703 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835728 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835730 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835721 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835690 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835722 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835723 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20835737 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P177213 - -
          Whole Mouse Genome G4122A A_51_P416927 BC119298.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P416927 BC119300.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P416927 NM_008763.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120065 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P177213 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00322974 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00322977 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00322979 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00322954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20835673 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835712 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835757 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00785744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835687 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835742 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835728 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835731 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835747 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835710 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00391951 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835711 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835719 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835746 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00322964 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835701 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00322955 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835720 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835761 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835751 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835708 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00294168 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835673 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00322962 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835682 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835725 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00322972 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835729 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00246965 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00732470 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835750 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835716 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835685 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835723 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00322968 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835707 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835759 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835764 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835694 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835703 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835715 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835738 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00054442 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835700 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835735 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00785748 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20835706 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10351858 - -
          Affymetrix HT MG-430 PM Array Plate 1422382_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10351857 - -
          Mouse Genome 430 2.0 Array 1422382_at - -
          Mouse Genome 430 2.0 Array 1422382_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18313_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18313_at - -
          Affymetrix Mouse Clariom S Array PSR0100013549.mm.1 NM_008763.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013549.mm.1 NM_008763.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013549.mm.1 TC0100001625.mm.1 MTA TC
          Mouse Expression Set 430 1422382_PM_at - -
          Mouse Expression Set 430 1422382_at - -
          Murine Genome U74v2 Set 102554_at - -
          Murine Genome U74v2 Set 102554_PM_at - -
          Mouse Genome 430A 2.0 Array 1422382_at - -
          Mouse Genome 430A 2.0 Array 1422382_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422382_at - -
          Affymetrix HT Mouse Genome 430A Array 1422382_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 603589 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6290494 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006290494 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1217706 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1217706 - -
          mouse-6 v1.1 (Illumina) scl14328.1.1_59-S - -
          Illumina Sentrix mouse-6.v1 bead array 6290494 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl14328.1.1_59-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6290494 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1217706 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1217706 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline