Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00493899_g1
          See other PHLDA2 GE Assays ›
          Gene Symbol:
          Phlda2
          Gene Name:
          pleckstrin homology-like domain, family A, member 2
          Gene Aliases:
          Ipl, Tssc3
          Chromosome Location:
          Chr.7: 143501545 - 143503145 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00493899_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Phlda2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009434.2 NP_033460.1 1 - 2 465 88
          GenBank mRNA AK131929.1 - 1 - 2 469 88
          BC019141.1 - 1 - 2 455 5042041 88
          CF615806.1 - 1 - 2 88
          CF617643.1 - 1 - 2 435 88
          Y15443.1 - 1 - 2 451 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004586.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Phlda2

          Gene Ontology Categories:

          Function(s)

          placenta development
          organ morphogenesis
          regulation of gene expression
          regulation of cell migration
          regulation of embryonic development
          regulation of glycogen metabolic process

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25601381 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25601378 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25601383 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25601380 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25601382 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25601372 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25601373 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25601371 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25601379 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25601377 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P350786 NM_009434.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P350786 AK131929.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P350786 BC019141.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P350786 CF615806.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P350786 CF617643.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P350786 NM_009434.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P350786 AK131929.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P350786 BC019141.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P350786 CF615806.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P350786 CF617643.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 700542 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137319 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P350786 NM_009434.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P350786 AK131929.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P350786 BC019141.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P350786 CF615806.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P350786 CF617643.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P350786 NM_009434.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P350786 AK131929.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P350786 BC019141.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P350786 CF615806.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P350786 CF617643.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P450623 NM_009434.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P450623 AK131929.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P450623 BC019141.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P450623 CF615806.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P450623 CF617643.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P350786 NM_009434.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P350786 AK131929.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P350786 BC019141.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P350786 CF615806.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P350786 CF617643.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P350786 NM_009434.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P350786 AK131929.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P350786 BC019141.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P350786 CF615806.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P350786 CF617643.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25601391 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25601372 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25601398 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25601378 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25601392 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25601381 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25601399 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25601364 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25601370 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25601366 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25601374 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25601394 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25601375 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25601383 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25601390 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25601376 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25601397 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25601388 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25601384 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25601368 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25601371 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25601385 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25601400 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P137319 - -
          Whole Mouse Genome G4122A A_51_P450623 NM_009434.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450623 AK131929.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450623 BC019141.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450623 CF615806.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450623 CF617643.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P350786 NM_009434.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P350786 AK131929.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P350786 BC019141.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P350786 CF615806.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P350786 CF617643.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P350786 NM_009434.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P350786 AK131929.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P350786 BC019141.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P350786 CF615806.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P350786 CF617643.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108132 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P137319 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05965154 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05965147 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06664391 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05965152 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05965142 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05965146 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05965148 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25601382 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25601381 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25601377 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25601383 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25601380 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25601371 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25601372 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25601373 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25601379 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25601378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25601417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25601370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25601400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25601376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25601391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25601429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25601384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25601402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25601413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25601392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25601412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25601365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25601364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25601371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25601374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25601388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25601385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25601381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25601433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25601424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25601404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25601425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25601366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25601421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25601405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25601399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25601415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25601430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25601375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25601378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25601383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25601410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25601387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25601419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06238016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25601405 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25601413 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25601402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25601370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25601378 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25601365 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25601400 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25601376 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25601391 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25601372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25601366 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06295028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25601392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25601394 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25601390 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25601368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25601388 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06664391 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25601375 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25601384 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25601404 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25601385 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06445380 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25601364 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25601397 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06454552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25601374 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25601398 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25601380 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06295025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25601412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25601387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05965152 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10569438 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569440 - -
          Affymetrix HT MG-430 PM Array Plate 1417837_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10569437 - -
          Mouse Genome 430 2.0 Array 1417837_PM_at - -
          Mouse Genome 430 2.0 Array 1417837_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22113_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22113_at - -
          Affymetrix Mouse Clariom S Array PSR0700038737.mm.1 NM_009434.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700038737.mm.1 NM_009434.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700038737.mm.1 TC0700004586.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700038738.mm.1 NM_009434.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700038738.mm.1 NM_009434.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700038738.mm.1 TC0700004586.mm.1 MTA TC
          Mouse Expression Set 430 1417837_PM_at - -
          Mouse Expression Set 430 1417837_at - -
          Murine Genome U74v2 Set 104548_PM_at - -
          Murine Genome U74v2 Set 104548_at - -
          Mouse Genome 430A 2.0 Array 1417837_PM_at - -
          Mouse Genome 430A 2.0 Array 1417837_at - -
          Affymetrix HT Mouse Genome 430A Array 1417837_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417837_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 762040 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4810494 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004810494 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2425029 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2425029 - -
          mouse-6 v1.1 (Illumina) scl30454.4.1_68-S - -
          Illumina Sentrix mouse-6.v1 bead array 4810494 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl30454.4.1_68-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4810494 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2425029 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2425029 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline