Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00495207_m1
          See other KRT17 GE Assays ›
          Gene Symbol:
          Krt17
          Gene Name:
          keratin 17
          Gene Aliases:
          K17, Krt1-17
          Chromosome Location:
          Chr.11: 100256217 - 100260989 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00495207_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Krt17
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010663.2 NP_034793.1 7 - 8 1235 81
          GenBank mRNA AB013608.1 - 7 - 8 1236 81
          BC032161.1 - 7 - 8 1282 81
          BC132454.1 - 7 - 8 1270 40130731 81
          BC132456.1 - 7 - 8 1270 40130733 81
          M13805.1 - 4 - 5 527 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100003763.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Krt17

          Gene Ontology Categories:

          Function(s) Process(es)

          morphogenesis of an epithelium
          signal transduction
          positive regulation of cell growth
          hair follicle morphogenesis
          keratinization
          intermediate filament organization
          positive regulation of translation
          positive regulation of hair follicle development
          structural molecule activity
          protein binding
          MHC class II receptor activity
          MHC class II protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1981994 M13805.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1981994 NM_010663.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1981994 AB013608.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 530567 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 400928 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 649439 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 135120 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P117360 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1981994 M13805.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1981994 NM_010663.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1981994 AB013608.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P433450 BC032161.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P433450 M13805.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P433450 NM_010663.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P433450 AB013608.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067090 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067072 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067083 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067088 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067085 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067079 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067091 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067077 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067060 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067087 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067073 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067082 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067084 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067061 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P117360 - -
          Whole Mouse Genome G4122A A_51_P433450 BC032161.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P433450 M13805.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P433450 NM_010663.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P433450 AB013608.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104996 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P117360 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09669412 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09669398 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09399163 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09399161 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09669404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09399169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09165259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09165265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09399179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09165264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067088 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09399174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09165248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067085 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10391072 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391070 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391068 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391069 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391071 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391074 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391067 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391073 - -
          Affymetrix HT MG-430 PM Array Plate 1420193_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1423227_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10391066 - -
          Mouse Genome 430 2.0 Array 1427841_at - -
          Mouse Genome 430 2.0 Array 1420193_at - -
          Mouse Genome 430 2.0 Array 1427841_PM_at - -
          Mouse Genome 430 2.0 Array 1423227_at - -
          Mouse Genome 430 2.0 Array 1420193_PM_at - -
          Mouse Genome 430 2.0 Array 1423227_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16667_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16667_at - -
          Affymetrix Mouse Clariom S Array PSR1100034532.mm.1 NM_010663.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034532.mm.1 TC1100003763.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034534.mm.1 NM_010663.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034534.mm.1 TC1100003763.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034533.mm.1 NM_010663.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034533.mm.1 TC1100003763.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034530.mm.1 NM_010663.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034530.mm.1 TC1100003763.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034531.mm.1 NM_010663.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034531.mm.1 TC1100003763.mm.1 MTA TC
          Mouse Expression Set 430 1427841_PM_at - -
          Mouse Expression Set 430 1420193_at - -
          Mouse Expression Set 430 1420193_PM_at - -
          Mouse Expression Set 430 1423227_PM_at - -
          Mouse Expression Set 430 1427841_at - -
          Mouse Expression Set 430 1423227_at - -
          Murine Genome U74v2 Set 92861_PM_i_at - -
          Murine Genome U74v2 Set 161496_PM_r_at - -
          Murine Genome U74v2 Set 161496_r_at - -
          Murine Genome U74v2 Set 92862_f_at - -
          Murine Genome U74v2 Set 92862_PM_f_at - -
          Murine Genome U74v2 Set 92861_i_at - -
          Mouse Genome 430A 2.0 Array 1423227_at - -
          Mouse Genome 430A 2.0 Array 1427841_PM_at - -
          Mouse Genome 430A 2.0 Array 1420193_PM_at - -
          Mouse Genome 430A 2.0 Array 1427841_at - -
          Mouse Genome 430A 2.0 Array 1423227_PM_at - -
          Mouse Genome 430A 2.0 Array 1420193_at - -
          Affymetrix HT Mouse Genome 430A Array 1423227_at - -
          Affymetrix HT Mouse Genome 430A Array 1423227_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420193_at - -
          Affymetrix HT Mouse Genome 430A Array 1420193_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 358579 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2230452 - -
          Illumina mouse-6 v1.0 expression beadchip 2230452 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002230452 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2661214 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2661214 - -
          mouse-6 v1.1 (Illumina) scl016667.1_175-S - -
          Illumina Sentrix mouse-6.v1 bead array 2230452 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl016667.1_175-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2230452 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2661214 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2661214 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline