Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00495683_m1
          See other TNFRSF17 GE Assays ›
          Gene Symbol:
          Tnfrsf17
          Gene Name:
          tumor necrosis factor receptor superfamily, member 17
          Gene Aliases:
          BCM, BCMA, Tnfrsf13, Tnfrsf13a
          Chromosome Location:
          Chr.16: 11313174 - 11320074 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00495683_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tnfrsf17
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011608.1 NP_035738.1 2 - 3 417 67
          GenBank mRNA AF061505.1 - 2 - 3 417 67
          AK020247.1 - 2 - 3 415 67
          BC027519.1 - 2 - 3 288 1349977 67

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tnfrsf17

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s)

          lymphocyte homeostasis
          immune system process

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P270807 AK020247.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P270807 BC027519.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P270807 NM_011608.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P270807 AF061505.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P270807 AK020247.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P270807 BC027519.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P270807 NM_011608.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P270807 AF061505.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 654791 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 968009 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 363122 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 866720 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 107032 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161587 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P270807 AK020247.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P270807 BC027519.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P270807 NM_011608.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P270807 AF061505.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P270807 AK020247.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P270807 BC027519.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P270807 NM_011608.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P270807 AF061505.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P270807 AK020247.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P270807 BC027519.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P270807 NM_011608.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P270807 AF061505.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P270807 AK020247.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P270807 BC027519.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P270807 NM_011608.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P270807 AF061505.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09550170 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09550167 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03821135 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03821134 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03821138 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03821132 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P161587 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09550158 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03821136 - -
          Whole Mouse Genome G4122A A_51_P270807 AK020247.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P270807 BC027519.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P270807 NM_011608.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P270807 AF061505.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P270807 AK020247.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P270807 BC027519.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P270807 NM_011608.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P270807 AF061505.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139751 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03821138 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P161587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30547241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30547252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30547240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30547245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30547250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30547246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30547253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30547243 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12554925 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949701 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949691 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949674 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949731 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949720 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949693 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949683 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949730 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949707 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949702 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949681 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949706 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30547250 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949703 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949713 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949676 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12554919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949697 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949690 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949700 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12701569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949673 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949698 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949711 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949715 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949695 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949668 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949672 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949708 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949705 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949669 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30547253 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30547256 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949689 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949710 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949725 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949712 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949732 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949679 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949719 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949716 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949696 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949728 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949722 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949714 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949671 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949729 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32949687 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10433585 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433586 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433587 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433588 - -
          Affymetrix HT MG-430 PM Array Plate 1420782_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10433584 - -
          Mouse Genome 430 2.0 Array 1420782_PM_at - -
          Mouse Genome 430 2.0 Array 1420782_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 21935_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 21935_PM_at - -
          Mouse Expression Set 430 1420782_at - -
          Mouse Expression Set 430 1420782_PM_at - -
          Murine Genome U74v2 Set 94190_at - -
          Murine Genome U74v2 Set 94190_PM_at - -
          Mouse Genome 430A 2.0 Array 1420782_PM_at - -
          Mouse Genome 430A 2.0 Array 1420782_at - -
          Affymetrix HT Mouse Genome 430A Array 1420782_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420782_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 754664 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5890408 - -
          Illumina mouse-6 v1.0 expression beadchip 5890408 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005890408 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1240522 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1240522 - -
          mouse-6 v1.1 (Illumina) scl49427.3.1_283-S - -
          Illumina Sentrix mouse-6.v1 bead array 5890408 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl49427.3.1_283-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5890408 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2854497 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2854497 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline