Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00498192_s1
          See other FKBPL GE Assays ›
          Gene Symbol:
          Fkbpl
          Gene Name:
          FK506 binding protein-like
          Gene Aliases:
          DIR1, NG7, Ppiase-X, Wisp39
          Chromosome Location:
          Chr.17: 34639574 - 34646327 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00498192_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fkbpl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019873.2 NP_063926.1 2 833 73
          GenBank mRNA AK139541.1 - 1 414 73
          BC046338.1 - 2 833 6395929 73

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fkbpl

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          protein peptidyl-prolyl isomerization
          chaperone-mediated protein folding
          peptidyl-prolyl cis-trans isomerase activity
          FK506 binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P515452 AK146018.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P515452 AK139541.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P515452 AK146018.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P515452 AK139541.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 261377 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P165752 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P515452 AK146018.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P515452 AK139541.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P515452 AK146018.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P515452 AK139541.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154996 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155001 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155002 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155007 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154993 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154998 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155000 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155013 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154992 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155026 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P165752 - -
          Whole Mouse Genome G4122A A_51_P515452 AK146018.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P515452 AK139541.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P515452 AK146018.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P515452 AK139541.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123142 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P165752 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13224117 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13224125 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13224115 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13224120 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13224124 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13224116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155038 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155000 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31154989 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13438129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13224116 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31154994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155015 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155001 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31154979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31154972 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31154977 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155012 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13530184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155013 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31154991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31154998 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31154974 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31154981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31154980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13291044 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31154982 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13503844 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31154997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13453614 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13291046 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31154969 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31154973 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31154985 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31154986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155014 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13291042 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31154966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13291045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13210785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13206335 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13251301 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10444438 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444437 - -
          Affymetrix HT MG-430 PM Array Plate 1422787_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10444436 - -
          Mouse Genome 430 2.0 Array 1422787_PM_at - -
          Mouse Genome 430 2.0 Array 1422787_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56299_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56299_at - -
          Mouse Expression Set 430 1422787_PM_at - -
          Mouse Expression Set 430 1422787_at - -
          Murine Genome U74v2 Set 102206_PM_at - -
          Murine Genome U74v2 Set 102206_at - -
          Mouse Genome 430A 2.0 Array 1422787_PM_at - -
          Mouse Genome 430A 2.0 Array 1422787_at - -
          Affymetrix HT Mouse Genome 430A Array 1422787_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422787_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 619946 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6550592 - -
          Illumina mouse-6 v1.0 expression beadchip 6550592 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006550592 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2710201 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2710201 - -
          mouse-6 v1.1 (Illumina) scl50813.2_82-S - -
          Illumina Sentrix mouse-6.v1 bead array 6550592 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50813.2_82-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6550592 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2933426 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2710201 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2933426 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2710201 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl50813.2_82-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline