Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00502775_m1
          See other HSBP1 GE Assays ›
          Gene Symbol:
          Hsbp1
          Gene Name:
          heat shock factor binding protein 1
          Gene Aliases:
          0610007A03Rik, AA408769, Hsp25
          Chromosome Location:
          Chr.8: 119344538 - 119348929 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00502775_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hsbp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024219.1 NP_077181.1 3 - 4 274 65
          GenBank mRNA AK010939.1 - 828 65
          AK018708.1 - 3 - 4 306 65
          AK050550.1 - 3 - 4 324 65
          AK078498.1 - 3 - 4 306 65
          AK145940.1 - 3 - 4 284 65
          BC002153.1 - 3 - 4 274 3485301 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001418.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hsbp1

          Gene Ontology Categories:

          Function(s) Process(es)

          muscle contraction
          endodermal cell differentiation
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26213863 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26213865 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26213861 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26213864 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26213862 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P544555 AK145940.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P544555 BC002153.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P544555 AK010939.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P544555 AK018708.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P544555 AK050550.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P544555 AK145940.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P544555 BC002153.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P544555 AK010939.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P544555 AK018708.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P544555 AK050550.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 587796 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 125929 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 664882 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 906912 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P143099 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P544555 AK145940.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P544555 BC002153.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P544555 AK010939.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P544555 AK018708.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P544555 AK050550.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P544555 AK145940.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P544555 BC002153.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P544555 AK010939.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P544555 AK018708.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P544555 AK050550.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P325624 AK145940.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P325624 BC002153.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P325624 AK050550.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P325624 NM_024219.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P325624 AK078498.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26213852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26213872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26213842 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26213875 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26213857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26213850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26213847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26213867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26213855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26213861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26213874 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26213849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26213862 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26213841 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26213864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26213843 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26213837 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26213839 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26213871 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26213845 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06661535 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P143099 - -
          Whole Mouse Genome G4122A A_52_P544556 AK145940.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P544556 BC002153.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P544556 AK010939.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P544556 AK018708.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P544556 AK050550.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P325624 AK145940.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P325624 BC002153.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P325624 AK050550.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P325624 NM_024219.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P325624 AK078498.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P544555 AK145940.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P544555 BC002153.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P544555 AK010939.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P544555 AK018708.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P544555 AK050550.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P544555 AK145940.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P544555 BC002153.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P544555 AK010939.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P544555 AK018708.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P544555 AK050550.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107926 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P143099 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07251253 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07251254 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07251251 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26213864 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26213862 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26213863 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26213865 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26213861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26213842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213916 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06920866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213913 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213909 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213845 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06884754 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213904 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06717890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213897 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213868 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06687934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06884749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213927 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213852 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07148946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213872 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07400347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07096473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06884755 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07400340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213898 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213857 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06768746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213906 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213908 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26213902 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10575864 - -
          Affymetrix Mouse Gene 1.0 ST Array 10575865 - -
          Affymetrix Mouse Gene 1.0 ST Array 10575863 - -
          Affymetrix Mouse Gene 1.0 ST Array 10575866 - -
          Affymetrix HT MG-430 PM Array Plate 1423906_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1451162_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10575861 - -
          Mouse Genome 430 2.0 Array 1451162_at - -
          Mouse Genome 430 2.0 Array 1423906_at - -
          Mouse Genome 430 2.0 Array 1451162_PM_at - -
          Mouse Genome 430 2.0 Array 1423906_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68196_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68196_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0800011508.mm.1 NM_024219.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800011508.mm.1 NM_024219.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800011508.mm.1 TC0800001418.mm.1 MTA TC
          Mouse Expression Set 430 1451162_at - -
          Mouse Expression Set 430 1423906_PM_at - -
          Mouse Expression Set 430 1423906_at - -
          Mouse Expression Set 430 1451162_PM_at - -
          Murine Genome U74v2 Set 93743_at - -
          Murine Genome U74v2 Set 93743_PM_at - -
          Mouse Genome 430A 2.0 Array 1423906_PM_at - -
          Mouse Genome 430A 2.0 Array 1451162_PM_at - -
          Mouse Genome 430A 2.0 Array 1451162_at - -
          Mouse Genome 430A 2.0 Array 1423906_at - -
          Affymetrix HT Mouse Genome 430A Array 1451162_at - -
          Affymetrix HT Mouse Genome 430A Array 1423906_at - -
          Affymetrix HT Mouse Genome 430A Array 1451162_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423906_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 306454 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6900687 - -
          Illumina mouse-6 v1.0 expression beadchip 103610301 - -
          Illumina mouse-6 v1.0 expression beadchip 6900687 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006900687 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2756369 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1229515 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2756369 - -
          mouse-6 v1.1 (Illumina) scl33261.5_41-S - -
          Illumina Sentrix mouse-6.v1 bead array 103610301 - -
          Illumina Sentrix mouse-6.v1 bead array 6900687 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl33261.5_41-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6900687 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103610301 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2756369 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2756369 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1229515 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl33261.5_41-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline