Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00503210_m1
          See other GET4 GE Assays ›
          Gene Symbol:
          Get4
          Gene Name:
          golgi to ER traffic protein 4 homolog (S. cerevisiae)
          Gene Aliases:
          1110007L15Rik, AW412535, Cee
          Chromosome Location:
          Chr.5: 139252324 - 139270050 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00503210_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Get4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001163316.1 NP_001156788.1 8 - 9 845 103
          NM_026269.2 NP_080545.2 8 - 9 1036 103
          GenBank mRNA AK003551.1 - 8 - 9 985 103
          AK207648.1 - 1 - 2 36 103
          BC019557.1 - 8 - 9 827 4163641 103

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500001669.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Get4

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          tail-anchored membrane protein insertion into ER membrane
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2210315 BC019557.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2210315 CJ048159.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2210315 NM_001163316.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2210315 NM_026269.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2210315 AK003551.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1969352 NM_001163316.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1969352 NM_026269.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1969352 AK003551.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 186502 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 378509 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 2961 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 453265 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 282825 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 1449 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 613171 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 474587 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 28507 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P108295 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P177684 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2210315 BC019557.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2210315 CJ048159.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2210315 NM_001163316.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2210315 NM_026269.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2210315 AK003551.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1969352 NM_001163316.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1969352 NM_026269.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1969352 AK003551.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24134461 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24134458 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24134453 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24134456 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24134451 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24134455 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24134443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24134457 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24134438 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24134452 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24134444 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24134463 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24134449 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24134460 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24134466 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24134439 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24134441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24134472 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24134468 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24134464 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24134447 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24134471 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24134470 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P108295 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05764856 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05764844 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P177684 - -
          Whole Mouse Genome G4122A A_51_P111212 NM_001163316.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P111212 NM_026269.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P111212 AK003551.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P488200 AK008315.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P488200 BC019557.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P488200 NM_001163316.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P488200 NM_026269.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P488200 AK003551.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105907 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P177684 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P108295 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04147330 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04147329 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04147327 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04183213 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04147315 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04147320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24134464 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24134476 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24134455 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04465765 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04183214 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04781437 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24134447 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04714438 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24134463 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24134458 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04147321 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04132197 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24134486 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24134457 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04949431 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04949424 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24134483 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24134493 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04949429 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04854522 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04781434 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24134439 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24134436 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24134494 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24134466 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24134468 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04218290 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04147318 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04398845 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24134444 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24134443 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04147329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04147327 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24134472 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24134478 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24134449 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24134484 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24134461 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24134492 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24134491 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04960758 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10526930 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526925 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526928 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526926 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526929 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526927 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526924 - -
          Affymetrix HT MG-430 PM Array Plate 1451700_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10526923 - -
          Mouse Genome 430 2.0 Array 1428528_PM_at - -
          Mouse Genome 430 2.0 Array 1451700_PM_a_at - -
          Mouse Genome 430 2.0 Array 1428528_at - -
          Mouse Genome 430 2.0 Array 1451700_a_at - -
          Affymetrix Mouse Clariom S Array PSR0500015414.mm.1 TC0500001669.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015414.mm.1 NM_001163316.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015414.mm.1 NM_001163316.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015414.mm.1 NM_026269.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015414.mm.1 NM_026269.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015417.mm.1 TC0500001669.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015417.mm.1 NM_001163316.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015417.mm.1 NM_001163316.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015417.mm.1 NM_026269.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015417.mm.1 NM_026269.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015416.mm.1 TC0500001669.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015416.mm.1 NM_001163316.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015416.mm.1 NM_001163316.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015416.mm.1 NM_026269.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015416.mm.1 NM_026269.2 Refseq
          Mouse Expression Set 430 1428528_PM_at - -
          Mouse Expression Set 430 1428528_at - -
          Mouse Expression Set 430 1451700_PM_a_at - -
          Mouse Expression Set 430 1451700_a_at - -
          Murine Genome U74v2 Set 99447_at - -
          Murine Genome U74v2 Set 99447_PM_at - -
          Mouse Genome 430A 2.0 Array 1428528_PM_at - -
          Mouse Genome 430A 2.0 Array 1428528_at - -
          Mouse Genome 430A 2.0 Array 1451700_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1451700_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1451700_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1451700_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 311856 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3840008 - -
          MouseWG-6 v2 Expression BeadChip 2120484 - -
          MouseWG-6 v2 Expression BeadChip 4120278 - -
          Illumina mouse-6 v1.0 expression beadchip 4120278 - -
          Illumina mouse-6 v1.0 expression beadchip 2120484 - -
          Illumina mouse-6 v1.0 expression beadchip 3840008 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003840008 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004120278 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002120484 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2622829 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2745954 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2621472 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2745954 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2621472 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2622829 - -
          mouse-6 v1.1 (Illumina) scl27066.10_346-S - -
          mouse-6 v1.1 (Illumina) scl067604.1_6-S - -
          mouse-6 v1.1 (Illumina) scl0067604.1_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 3840008 - -
          Illumina Sentrix mouse-6.v1 bead array 4120278 - -
          Illumina Sentrix mouse-6.v1 bead array 2120484 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl067604.1_6-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl27066.10_346-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0067604.1_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4120278 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3840008 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2120484 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2621471 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2622829 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2621472 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2745954 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2622829 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2745954 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2621472 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2621471 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl067604.1_6-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl27066.10_346-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0067604.1_0-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline