Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00503537_m1
          See other CD177 GE Assays ›
          Gene Symbol:
          Cd177
          Gene Name:
          CD177 antigen
          Gene Aliases:
          1190003K14Rik, Pdp3
          Chromosome Location:
          Chr.7: 24743983 - 24760334 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00503537_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd177
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026862.3 NP_081138.2 16 - 17 2243 77
          GenBank mRNA AK004472.1 - 3 - 4 403 77
          BC005615.1 - 11 - 12 1613 77
          BC027283.1 - 16 - 17 2243 2648499 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002611.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cd177

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P385718 BC005615.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P385718 BC027283.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P385718 NM_026862.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P385718 BC005615.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P385718 BC027283.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P385718 NM_026862.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 767205 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 790656 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 955576 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 649854 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 966728 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 9566 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 8761 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 67900 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 118373 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100239 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P385718 BC005615.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P385718 BC027283.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P385718 NM_026862.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P385718 BC005615.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P385718 BC027283.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P385718 NM_026862.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006469 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006468 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006485 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006463 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006476 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006480 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006459 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006462 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006489 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006477 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006455 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006460 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006478 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006467 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006490 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006454 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006458 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006475 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006492 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006487 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006465 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006464 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006484 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P165063 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06141673 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01485801 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01485798 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100239 - -
          Whole Mouse Genome G4122A A_51_P385718 BC005615.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P385718 BC027283.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P385718 NM_026862.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P385718 BC005615.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P385718 BC027283.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P385718 NM_026862.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115880 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133008 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121303 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113040 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P165063 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06141663 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100239 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06100332 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06100334 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05930737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006390 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05930737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06215255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006495 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006507 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05937404 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05937406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05971313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006468 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05937407 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006463 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06215247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06542751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05914639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06270977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06481279 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06379233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05930742 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06530536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006446 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06215254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06542749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06270970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06481278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006480 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06379234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06379232 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006422 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006445 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05930738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06481285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05891790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05895651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06270974 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006441 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06270972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06100332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06542752 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05914640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006444 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06468359 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06619898 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06650349 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006442 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06514505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05914637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05930735 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05914634 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10560896 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560889 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560897 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560888 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560893 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560894 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560901 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560899 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560890 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560898 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560887 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560895 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560892 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560900 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560891 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560902 - -
          Affymetrix HT MG-430 PM Array Plate 1424509_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10560886 - -
          Mouse Genome 430 2.0 Array 1424509_PM_at - -
          Mouse Genome 430 2.0 Array 1424509_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68891_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68891_at - -
          Affymetrix Mouse Clariom S Array PSR0700022510.mm.1 TC0700002611.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700022510.mm.1 NM_026862.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700022510.mm.1 NM_026862.3 Refseq
          Mouse Expression Set 430 1424509_at - -
          Mouse Expression Set 430 1424509_PM_at - -
          Murine Genome U74v2 Set 165417_PM_at - -
          Murine Genome U74v2 Set 165417_at - -
          Mouse Genome 430A 2.0 Array 1424509_at - -
          Mouse Genome 430A 2.0 Array 1424509_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424509_at - -
          Affymetrix HT Mouse Genome 430A Array 1424509_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 721197 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 670035 - -
          Illumina mouse-6 v1.0 expression beadchip 670035 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000670035 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1225233 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1225233 - -
          mouse-6 v1.1 (Illumina) scl068891.3_33-S - -
          Illumina Sentrix mouse-6.v1 bead array 670035 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl068891.3_33-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 670035 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2789900 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1225233 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1225233 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2789900 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline