Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00504878_m1
          See other CYP2A12 GE Assays ›
          Gene Symbol:
          Cyp2a12
          Gene Name:
          cytochrome P450, family 2, subfamily a, polypeptide 12
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 27029090 - 27036818 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00504878_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyp2a12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133657.1 NP_598418.1 4 - 5 666 79
          GenBank mRNA BC018356.1 - 4 - 5 666 4195640 79
          BC094017.1 - 4 - 5 654 5101743 79
          L06463.1 - 4 - 5 652 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000446.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection P450 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cyp2a12

          Panther Classification:

          Molecular Function -

          oxidoreductase oxygenase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          xenobiotic metabolic process
          coumarin metabolic process
          epoxygenase P450 pathway
          exogenous drug catabolic process
          oxidation-reduction process
          monooxygenase activity
          iron ion binding
          arachidonic acid epoxygenase activity
          steroid hydroxylase activity
          oxidoreductase activity
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced flavin or flavoprotein as one donor, and incorporation of one atom of oxygen
          oxygen binding
          heme binding
          metal ion binding
          aromatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2126333 L06463.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2126333 NM_133657.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2126333 BC018356.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2126333 BC094017.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 587533 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 167261 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2126333 L06463.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2126333 NM_133657.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2126333 BC018356.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2126333 BC094017.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P206736 NM_133657.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P206736 BC018356.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P206736 BC094017.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25015343 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25015348 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25015342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25015326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25015333 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01488134 - -
          Whole Mouse Genome G4122A A_51_P206736 NM_133657.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P206736 BC018356.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P206736 BC094017.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132379 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111933 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01488134 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06347626 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06546641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25015358 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015371 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015362 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015337 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015384 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015376 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015358 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015381 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06354693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015378 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015348 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06518190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015418 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015353 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25015343 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10551289 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551288 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551291 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551290 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551292 - -
          Affymetrix HT MG-430 PM Array Plate 1418821_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10551287 - -
          Mouse Genome 430 2.0 Array 1418821_PM_at - -
          Mouse Genome 430 2.0 Array 1418821_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13085_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13085_PM_at - -
          Affymetrix Murine Genome 6500A Array Msa11570_at - -
          Affymetrix Mouse Clariom S Array PSR0700003501.mm.1 TC0700000446.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700003501.mm.1 NM_133657.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003501.mm.1 NM_133657.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003505.mm.1 TC0700000446.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700003505.mm.1 NM_133657.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003505.mm.1 NM_133657.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003500.mm.1 TC0700000446.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700003500.mm.1 NM_133657.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003500.mm.1 NM_133657.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003502.mm.1 TC0700000446.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700003502.mm.1 NM_133657.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003502.mm.1 NM_133657.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003499.mm.1 TC0700000446.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700003499.mm.1 NM_133657.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003499.mm.1 NM_133657.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003503.mm.1 TC0700000446.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700003503.mm.1 NM_133657.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003503.mm.1 NM_133657.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003504.mm.1 TC0700000446.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700003504.mm.1 NM_133657.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003504.mm.1 NM_133657.1 Refseq
          Mouse Expression Set 430 1418821_at - -
          Mouse Expression Set 430 1418821_PM_at - -
          Murine Genome U74v2 Set 103670_PM_at - -
          Murine Genome U74v2 Set 103670_at - -
          Mouse Genome 430A 2.0 Array 1418821_PM_at - -
          Mouse Genome 430A 2.0 Array 1418821_at - -
          Affymetrix HT Mouse Genome 430A Array 1418821_at - -
          Affymetrix HT Mouse Genome 430A Array 1418821_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 855489 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3360619 - -
          Illumina mouse-6 v1.0 expression beadchip 1090487 - -
          Illumina mouse-6 v1.0 expression beadchip 3360619 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003360619 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001090487 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2645815 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2645815 - -
          mouse-6 v1.1 (Illumina) scl0013085.1_15-S - -
          mouse-6 v1.1 (Illumina) scl013085.8_30-S - -
          Illumina Sentrix mouse-6.v1 bead array 1090487 - -
          Illumina Sentrix mouse-6.v1 bead array 3360619 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0013085.1_15-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl013085.8_30-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1090487 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3360619 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2645815 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2645815 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline