Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00507118_m1
          See other PLXNB2 GE Assays ›
          Gene Symbol:
          Plxnb2
          Gene Name:
          plexin B2
          Gene Aliases:
          Debt, plexin-B2
          Chromosome Location:
          Chr.15: 89155546 - 89181140 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00507118_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Plxnb2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001159521.2 NP_001152993.1 9 - 10 2041 86
          NM_001284506.1 NP_001271435.1 8 - 9 1955 86
          NM_138749.3 NP_620088.2 9 - 10 2005 86
          GenBank mRNA AB093229.1 - 5 - 6 589 86
          AK147538.1 - 9 - 10 2006 86
          AK154627.1 - 9 - 10 1956 86
          BC007481.1 - 6 - 7 803 86
          BC031202.1 - 9 - 10 1925 86
          BC157960.1 - 9 - 10 1976 9088132 86
          BC157961.1 - 9 - 10 1976 9088133 86
          BC158086.1 - 9 - 10 1942 9088174 86
          BC172162.1 - 9 - 10 1942 9054832 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500002040.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the BC007481.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Plxnb2

          Gene Ontology Categories:

          Function(s) Process(es)

          neural tube closure
          regulation of protein phosphorylation
          signal transduction
          multicellular organism development
          neuroblast proliferation
          brain development
          regulation of cell shape
          regulation of GTPase activity
          positive regulation of axonogenesis
          semaphorin-plexin signaling pathway
          regulation of neuron migration
          protein binding
          semaphorin receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1967266 BC003293.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1967266 BC007481.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1967266 BC036274.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1967266 NM_001159521.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1967266 NM_138749.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 532479 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 524730 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 963113 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 663727 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 792632 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 482862 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 29922 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 580291 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 23762 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 904504 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 880777 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 453386 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 533800 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 639401 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 582271 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 957952 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 12407 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 21318 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 79883 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P151233 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P127105 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136286 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1967266 BC003293.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1967266 BC007481.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1967266 BC036274.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1967266 NM_001159521.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1967266 NM_138749.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P311068 BC007481.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P311068 BC031202.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P311068 BC157960.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P311068 BC157961.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P311068 BC158086.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P136286 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08454697 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P127105 - -
          Whole Mouse Genome G4122A A_52_P311068 BC007481.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P311068 BC031202.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P311068 BC157960.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P311068 BC157961.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P311068 BC158086.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P678942 BC003293.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P678942 BC007481.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P678942 BC036274.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P678942 AB093229.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P678942 NM_001159521.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138176 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P136286 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P127105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423570 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12186244 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423650 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423614 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12230997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423643 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423655 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12186264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423637 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423612 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423621 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12118448 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423646 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12255836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423616 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12230969 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12255839 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12186282 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12230980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12230986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423606 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423619 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423572 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423642 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12118456 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423610 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423566 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30423628 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10431448 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431431 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431456 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431451 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431426 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431444 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431442 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431439 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431452 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431462 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431445 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431458 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431427 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431459 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431457 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431432 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431430 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431449 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431437 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431429 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431436 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431434 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431443 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431440 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431455 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431428 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431435 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431450 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431425 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431454 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431433 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431460 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431461 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431453 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431441 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431446 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431438 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431447 - -
          Affymetrix HT MG-430 PM Array Plate 1416683_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1438638_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10431424 - -
          Mouse Genome 430 2.0 Array 1416683_PM_at - -
          Mouse Genome 430 2.0 Array 1416683_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 140570_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 140570_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1500016322.mm.1 NM_001159521.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016322.mm.1 NM_001284506.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016322.mm.1 NM_138749.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016322.mm.1 TC1500002040.mm.1 MTA TC
          Mouse Expression Set 430 1416683_PM_at - -
          Mouse Expression Set 430 1416683_at - -
          Murine Genome U74v2 Set 160311_at - -
          Murine Genome U74v2 Set 96464_at - -
          Murine Genome U74v2 Set 160311_PM_at - -
          Murine Genome U74v2 Set 96464_PM_at - -
          Mouse Genome 430A 2.0 Array 1416683_PM_at - -
          Mouse Genome 430A 2.0 Array 1416683_at - -
          Affymetrix HT Mouse Genome 430A Array 1416683_at - -
          Affymetrix HT Mouse Genome 430A Array 1416683_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 344972 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106020176 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2461098 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline