Search
Search

Ccdc53| Gene Symbol: Ccdc53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122960.1 | NP_001116432.1 | 3 - 4 | 364 | 95 | |
| NM_026070.3 | NP_080346.1 | 3 - 4 | 364 | 95 | ||
| NM_027487.1 | NP_081763.1 | 3 - 4 | 364 | 95 | ||
| GenBank mRNA | AK004206.1 | - | 3 - 4 | 279 | 95 | |
| AK009358.1 | - | 3 - 4 | 260 | 95 | ||
| AK013835.1 | - | 3 - 4 | 369 | 95 | ||
| AK017732.1 | - | 3 - 4 | 292 | 95 | ||
| AK158822.1 | - | 3 - 4 | 365 | 95 | ||
| BC029256.1 | - | 3 - 4 | 261 | 3661472 | 95 | |
| CK795584.1 | - | 3 - 4 | 316 | 95 | ||
Set Membership: |
|