Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00509903_g1
          See other HAGHL GE Assays ›
          Gene Symbol:
          Haghl
          Gene Name:
          hydroxyacylglutathione hydrolase-like
          Gene Aliases:
          1500017E18Rik, 2810014I23Rik, C330022E15Rik
          Chromosome Location:
          Chr.17: 25779843 - 25785669 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00509903_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Haghl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001271433.1 NP_001258362.1 4 - 5 530 56
          NM_001271434.1 NP_001258363.1 4 - 5 530 56
          NM_001271435.1 NP_001258364.1 4 - 5 530 56
          NM_001271436.1 NP_001258365.1 3 - 4 344 56
          NM_001271437.1 NP_001258366.1 3 - 4 339 56
          NM_026897.3 NP_081173.1 3 - 4 515 56
          GenBank mRNA AK005274.1 - 3 - 4 511 56
          AK012748.1 - 4 - 5 527 56
          AK021220.1 - 3 - 4 341 56
          AK034798.1 - 4 - 5 505 56
          AK047517.1 - 4 - 5 485 56
          AK154248.1 - 3 - 4 320 56
          BC005586.1 - 4 - 5 476 56
          BC030466.1 - 4 - 5 515 5253213 56
          BC083322.1 - 4 - 5 469 6820885 56

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700001672.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Haghl

          Gene Ontology Categories:

          Function(s) Process(es)

          glutathione biosynthetic process
          biological_process
          molecular_function
          hydroxyacylglutathione hydrolase activity
          zinc ion binding
          CoA hydrolase activity
          hydrolase activity
          2-oxoglutaryl-CoA thioesterase activity
          2,4,4-trimethyl-3-oxopentanoyl-CoA thioesterase activity
          3-isopropylbut-3-enoyl-CoA thioesterase activity
          glutaryl-CoA hydrolase activity
          metal ion binding
          acyl-CoA hydrolase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1700015284.mm.1 NM_001271433.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700015284.mm.1 NM_001271434.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700015284.mm.1 NM_001271435.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700015284.mm.1 NM_001271436.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700015284.mm.1 NM_001271437.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700015284.mm.1 NM_026897.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700015284.mm.1 TC1700001672.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700015285.mm.1 NM_001271433.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700015285.mm.1 NM_001271434.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700015285.mm.1 NM_001271435.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700015285.mm.1 NM_001271436.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700015285.mm.1 NM_001271437.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700015285.mm.1 NM_026897.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700015285.mm.1 TC1700001672.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline