Search
Search

Carkd| Gene Symbol: Carkd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190357.1 | NP_001177286.1 | 7 - 8 | 673 | 64 | |
| NM_001293661.1 | NP_001280590.1 | 7 - 8 | 673 | 64 | ||
| NM_026995.4 | NP_081271.2 | 7 - 8 | 725 | 64 | ||
| GenBank mRNA | AK009254.1 | - | 7 - 8 | 606 | 64 | |
| AK012456.1 | - | 7 - 8 | 613 | 64 | ||
| AK013028.1 | - | 7 - 8 | 694 | 64 | ||
| AK013069.1 | - | 7 - 8 | 592 | 64 | ||
| AK013420.1 | - | 7 - 8 | 597 | 64 | ||
| AK154142.1 | - | 7 - 8 | 707 | 64 | ||
| AK171034.1 | - | 7 - 8 | 707 | 64 | ||
| BC019538.1 | - | 7 - 8 | 645 | 4211749 | 64 | |
| BC021955.1 | - | 7 - 8 | 586 | 5148177 | 64 | |
| DV045296.1 | - | 7 - 8 | 611 | 64 | ||
Set Membership: |
|