Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00510269_m1
          See other 2010107G23RIK GE Assays ›
          Gene Symbol:
          2010107G23Rik
          Gene Name:
          RIKEN cDNA 2010107G23 gene
          Gene Aliases:
          AA545165
          Chromosome Location:
          Chr.10: 62107656 - 62111074 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00510269_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2010107G23Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027251.3 NP_081527.1 3 - 4 266 55
          GenBank mRNA AK008338.1 - 2 - 3 186 55
          AK035688.1 - 3 - 4 267 55

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000002252.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 2010107G23Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27184272 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27184273 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27184271 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27184269 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27184270 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P308029 BC024943.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P308029 BC024943.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 529146 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 717346 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 118832 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137309 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P308029 BC024943.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P308029 BC024943.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27184291 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27184261 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27184296 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27184259 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27184277 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27184293 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27184280 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27184271 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27184272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27184276 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27184266 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27184284 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27184268 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27184287 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27184285 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27184263 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27184282 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27184275 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27184295 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27184273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27184262 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27184260 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27184255 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27184269 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27184279 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27184289 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P137309 - -
          Whole Mouse Genome G4122A A_51_P308029 BC024943.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P308029 BC024943.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P232935 BC024943.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P232935 NM_027251.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P232935 AK008338.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P232935 AK035688.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115294 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P137309 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08718235 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08718231 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08396701 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08396700 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08296054 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08880678 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08396698 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08296051 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08718234 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27184271 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27184269 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27184272 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27184273 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27184270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27184242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184223 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184232 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08919441 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08279945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08500082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184275 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08385154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08919443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08496256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08718234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08274880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08496248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08500089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184230 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08627076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184202 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08279947 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184269 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08274878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08880678 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184280 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08281350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08385146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184268 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08592893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08509060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27184248 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10369526 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369530 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369529 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369527 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369528 - -
          Affymetrix HT MG-430 PM Array Plate 1424072_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10369525 - -
          Mouse Genome 430 2.0 Array 1424072_at - -
          Mouse Genome 430 2.0 Array 1424072_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69894_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69894_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1000015958.mm.1 NM_027251.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015958.mm.1 NM_027251.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015958.mm.1 TC1000002252.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000015966.mm.1 NM_027251.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015966.mm.1 NM_027251.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015966.mm.1 TC1000002252.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000015960.mm.1 NM_027251.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015960.mm.1 NM_027251.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015960.mm.1 TC1000002252.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000015976.mm.1 NM_027251.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015976.mm.1 NM_027251.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015976.mm.1 TC1000002252.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000015964.mm.1 NM_027251.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015964.mm.1 NM_027251.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015964.mm.1 TC1000002252.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000015965.mm.1 NM_027251.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015965.mm.1 NM_027251.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015965.mm.1 TC1000002252.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000015961.mm.1 NM_027251.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015961.mm.1 NM_027251.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015961.mm.1 TC1000002252.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000015969.mm.1 NM_027251.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015969.mm.1 NM_027251.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015969.mm.1 TC1000002252.mm.1 MTA TC
          Mouse Expression Set 430 1424072_PM_at - -
          Mouse Expression Set 430 1424072_at - -
          Mouse Genome 430A 2.0 Array 1424072_at - -
          Mouse Genome 430A 2.0 Array 1424072_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424072_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424072_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 925815 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 940288 - -
          Illumina mouse-6 v1.0 expression beadchip 940288 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000940288 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1240888 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1240888 - -
          mouse-6 v1.1 (Illumina) scl37906.4_416-S - -
          Illumina Sentrix mouse-6.v1 bead array 940288 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37906.4_416-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 940288 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2803564 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2803564 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline