Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00510679_m1
          See other SEPT12 GE Assays ›
          Gene Symbol:
          Sept12
          Gene Name:
          septin 12
          Gene Aliases:
          1700028G04Rik, 4933413B09Rik, Septin12
          Chromosome Location:
          Chr.16: 4986858 - 4997852 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00510679_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sept12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027669.3 NP_081945.1 7 - 8 859 75
          NM_029374.1 NP_083650.1 7 - 8 859 75
          GenBank mRNA AK006458.1 - 7 - 8 797 75
          AK016798.1 - 7 - 8 787 75
          AK077091.1 - 7 - 8 860 75
          BC115618.1 - 7 - 8 770 40102763 75
          BC115619.1 - 6 - 7 905 40102765 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600001149.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sept12

          Panther Classification:

          Molecular Function -

          cytoskeletal protein

          Gene Ontology Categories:

          Process(es)

          GTP binding
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P453196 BC115618.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P453196 BC115619.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P453196 AK006458.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P453196 AK016798.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P453196 AK077091.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P453196 BC115618.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P453196 BC115619.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P453196 AK006458.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P453196 AK016798.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P453196 AK077091.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 869314 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 754046 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 719568 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 622069 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 490391 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 938291 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 13352 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 67085 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152011 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100687 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P453196 BC115618.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P453196 BC115619.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P453196 AK006458.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P453196 AK016798.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P453196 AK077091.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P453196 BC115618.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P453196 BC115619.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P453196 AK006458.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P453196 AK016798.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P453196 AK077091.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30513155 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30513144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30513134 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30513137 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30513139 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30513141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30513153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30513147 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30513150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30513152 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P152011 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100687 - -
          Whole Mouse Genome G4122A A_51_P453196 BC115618.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P453196 BC115619.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P453196 AK006458.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P453196 AK016798.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P453196 AK077091.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P453196 BC115618.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P453196 BC115619.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P453196 AK006458.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P453196 AK016798.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P453196 AK077091.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126463 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P152011 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100687 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12982136 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12982145 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12982135 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12982138 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12982140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30513057 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30513135 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30513134 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30513139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30513137 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12883325 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30513145 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30513101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30513121 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30513123 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30513099 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30513100 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30513142 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30513096 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30513150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30513097 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30513128 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12981734 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12550784 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12921374 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30513141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12736117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30513164 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30513111 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30513113 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12550783 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30513102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12971023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12778528 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30513172 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12524703 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30513167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30513125 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12735657 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30513104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30513122 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30513168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30513152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30513103 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30513147 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12550785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12573432 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30513129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12982146 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30513094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30513133 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30513119 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30513095 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30513126 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13014345 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12882813 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10437476 - -
          Affymetrix Mouse Gene 1.0 ST Array 10437481 - -
          Affymetrix Mouse Gene 1.0 ST Array 10437475 - -
          Affymetrix Mouse Gene 1.0 ST Array 10437474 - -
          Affymetrix Mouse Gene 1.0 ST Array 10437477 - -
          Affymetrix Mouse Gene 1.0 ST Array 10437480 - -
          Affymetrix Mouse Gene 1.0 ST Array 10437478 - -
          Affymetrix Mouse Gene 1.0 ST Array 10437479 - -
          Affymetrix HT MG-430 PM Array Plate 1438298_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1460497_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10437472 - -
          Mouse Genome 430 2.0 Array 1438298_a_at - -
          Mouse Genome 430 2.0 Array 1460497_PM_a_at - -
          Mouse Genome 430 2.0 Array 1438298_PM_a_at - -
          Mouse Genome 430 2.0 Array 1460497_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71089_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71089_at - -
          Affymetrix Mouse Clariom S Array PSR1600009161.mm.1 NM_027669.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600009161.mm.1 NM_027669.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600009161.mm.1 TC1600001149.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600009165.mm.1 NM_027669.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600009165.mm.1 NM_027669.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600009165.mm.1 NM_029374.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600009165.mm.1 NM_029374.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600009165.mm.1 TC1600001149.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600009164.mm.1 NM_027669.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600009164.mm.1 NM_027669.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600009164.mm.1 TC1600001149.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600009163.mm.1 NM_027669.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600009163.mm.1 NM_027669.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600009163.mm.1 TC1600001149.mm.1 MTA TC
          Mouse Expression Set 430 1438298_a_at - -
          Mouse Expression Set 430 1438298_PM_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1438298_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1438298_PM_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1460497_PM_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1460497_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 611586 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102340538 - -
          MouseWG-6 v2 Expression BeadChip 3290487 - -
          Illumina mouse-6 v1.0 expression beadchip 102340538 - -
          Illumina mouse-6 v1.0 expression beadchip 3290487 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003290487 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1236861 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1236861 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1224756 - -
          mouse-6 v1.1 (Illumina) scl0071089.2_105-S - -
          mouse-6 v1.1 (Illumina) scl48795.8_120-S - -
          Illumina Sentrix mouse-6.v1 bead array 3290487 - -
          Illumina Sentrix mouse-6.v1 bead array 102340538 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0071089.2_105-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3290487 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102340538 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1236861 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1236861 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-8pzg4:80/100.66.75.138:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline