Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00511099_m1
          See other SLC25A35 GE Assays ›
          Gene Symbol:
          Slc25a35
          Gene Name:
          solute carrier family 25, member 35
          Gene Aliases:
          1810012H11Rik
          Chromosome Location:
          Chr.11: 68968108 - 68972516 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00511099_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc25a35
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028048.2 NP_082324.1 1 - 2 902 122
          GenBank mRNA AK007453.1 - 1 - 2 904 122
          AK035739.1 - 885 122
          AK157873.1 - 1 - 2 896 122
          AK171131.1 - 1 - 2 909 122
          BC004569.1 - 1 - 2 342 122
          BC019996.1 - 1 - 2 844 4207271 122

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000887.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc25a35

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27892299 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27892300 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27892295 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27892298 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27892297 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27892296 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P436447 BC004569.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P436447 BC019996.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P436447 AK035739.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P436447 NM_028048.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P436447 AK007453.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P436447 BC004569.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P436447 BC019996.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P436447 AK035739.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P436447 NM_028048.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P436447 AK007453.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 666323 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 265210 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 188873 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 177174 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P436447 BC004569.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P436447 BC019996.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P436447 AK035739.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P436447 NM_028048.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P436447 AK007453.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P436447 BC004569.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P436447 BC019996.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P436447 AK035739.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P436447 NM_028048.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P436447 AK007453.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27892324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27892326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27892313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27892328 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27892298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27892302 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27892294 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27892310 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27892327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27892296 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27892297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27892291 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27892321 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27892292 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27892320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27892301 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27892305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27892329 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27892290 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27892293 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27892306 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27892308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27892300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27892312 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27892295 - -
          Whole Mouse Genome G4122A A_52_P436447 BC004569.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P436447 BC019996.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P436447 AK035739.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P436447 NM_028048.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P436447 AK007453.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P436447 BC004569.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P436447 BC019996.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P436447 AK035739.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P436447 NM_028048.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P436447 AK007453.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P590756 AK035739.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09360233 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09360237 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09360232 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09360217 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09360225 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09271019 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27892297 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27892295 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27892299 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27892296 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27892300 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27892298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27892297 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27892321 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09360217 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27892317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27892327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27892308 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27892304 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27892311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09360233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27892282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27892310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09360221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09360228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27892298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27892313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27892306 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27892323 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27892329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27892315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27892328 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27892318 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27892291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27892312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27892290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27892285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27892288 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27892284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27892286 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10377374 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377377 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377375 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377373 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377378 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377376 - -
          Affymetrix HT MG-430 PM Array Plate 1451489_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10377372 - -
          Mouse Genome 430 2.0 Array 1451489_at - -
          Mouse Genome 430 2.0 Array 1451489_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71998_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71998_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100007413.mm.1 NM_028048.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007413.mm.1 NM_028048.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007413.mm.1 TC1100000887.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100007409.mm.1 NM_028048.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007409.mm.1 NM_028048.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007409.mm.1 TC1100000887.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100007411.mm.1 NM_028048.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007411.mm.1 NM_028048.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007411.mm.1 TC1100000887.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100007412.mm.1 NM_028048.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007412.mm.1 NM_028048.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007412.mm.1 TC1100000887.mm.1 MTA TC
          Mouse Expression Set 430 1451489_at - -
          Mouse Expression Set 430 1451489_PM_at - -
          Mouse Genome 430A 2.0 Array 1451489_at - -
          Mouse Genome 430A 2.0 Array 1451489_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451489_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451489_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 896093 - -
          ABI Mouse Genome Survey Microarray 914378 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4590341 - -
          Illumina mouse-6 v1.0 expression beadchip 106860524 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004590341 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1258728 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1258728 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1219114 - -
          mouse-6 v1.1 (Illumina) scl0071998.2_125-S - -
          mouse-6 v1.1 (Illumina) scl41385.5_416-S - -
          Illumina Sentrix mouse-6.v1 bead array 4590341 - -
          Illumina Sentrix mouse-6.v1 bead array 106860524 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0071998.2_125-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106860524 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4590341 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1258728 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1258728 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline