Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00511986_m1
          See other TMEM43 GE Assays ›
          Gene Symbol:
          Tmem43
          Gene Name:
          transmembrane protein 43
          Gene Aliases:
          1200015A22Rik, LUMA
          Chromosome Location:
          Chr.6: 91473751 - 91488458 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00511986_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem43
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028766.2 NP_083042.1 7 - 8 718 72
          GenBank mRNA AK004778.1 - 7 - 8 719 72
          AK037165.1 - 7 - 8 689 71
          AK050173.1 - 7 - 8 757 72
          AK151693.1 - 7 - 8 711 72
          AK151766.1 - 7 - 8 736 72
          AK152078.1 - 7 - 8 711 72
          AK168559.1 - 7 - 8 726 72
          BC005521.1 - 7 - 8 72
          BC024933.1 - 7 - 8 3990353 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600001087.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem43

          Gene Ontology Categories:

          Function(s) Process(es)

          nuclear membrane organization
          protein self-association

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24639211 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24639212 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P136998 AK004778.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P136998 NM_028766.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 867928 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 401790 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 102328 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 885102 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 921066 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 365625 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 638847 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 121226 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 739723 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 411191 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 96329 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P133643 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156470 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P136998 AK004778.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P136998 NM_028766.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639225 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639213 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639243 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639221 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639249 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639224 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639211 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639212 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639232 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639215 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639247 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639241 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639222 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639219 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24639250 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P133643 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05981081 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05981097 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P156470 - -
          Whole Mouse Genome G4122A A_51_P517157 BC024933.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P517157 AK168559.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P517157 AK152078.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P517157 AK050173.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P517157 AK151693.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136998 AK004778.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136998 NM_028766.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P133643 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P156470 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05740631 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05740641 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05740639 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24639212 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24639211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24639251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639269 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05471032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05570922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639336 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639214 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05839690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639219 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05839679 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639266 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639232 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05839694 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05740644 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639333 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639220 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05515906 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639230 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05515905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05168143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05839681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05168140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05570953 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639231 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05570926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24639237 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10540115 - -
          Affymetrix Mouse Gene 1.0 ST Array 10540106 - -
          Affymetrix Mouse Gene 1.0 ST Array 10540110 - -
          Affymetrix Mouse Gene 1.0 ST Array 10540108 - -
          Affymetrix Mouse Gene 1.0 ST Array 10540111 - -
          Affymetrix Mouse Gene 1.0 ST Array 10540114 - -
          Affymetrix Mouse Gene 1.0 ST Array 10540116 - -
          Affymetrix Mouse Gene 1.0 ST Array 10540107 - -
          Affymetrix Mouse Gene 1.0 ST Array 10540113 - -
          Affymetrix Mouse Gene 1.0 ST Array 10540109 - -
          Affymetrix Mouse Gene 1.0 ST Array 10540112 - -
          Affymetrix Mouse Gene 1.0 ST Array 10540117 - -
          Affymetrix HT MG-430 PM Array Plate 1439418_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1426434_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10540105 - -
          Mouse Genome 430 2.0 Array 1439418_x_at - -
          Mouse Genome 430 2.0 Array 1426434_at - -
          Mouse Genome 430 2.0 Array 1426434_PM_at - -
          Mouse Genome 430 2.0 Array 1439418_PM_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74122_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74122_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600008507.mm.1 TC0600001087.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600008507.mm.1 NM_028766.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600008507.mm.1 NM_028766.2 Refseq
          Mouse Expression Set 430 1439418_PM_x_at - -
          Mouse Expression Set 430 1439418_x_at - -
          Mouse Expression Set 430 1426434_PM_at - -
          Mouse Expression Set 430 1426434_at - -
          Murine Genome U74v2 Set 98633_PM_at - -
          Murine Genome U74v2 Set 98633_at - -
          Mouse Genome 430A 2.0 Array 1439418_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1439418_x_at - -
          Mouse Genome 430A 2.0 Array 1426434_at - -
          Mouse Genome 430A 2.0 Array 1426434_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1439418_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1439418_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1426434_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1426434_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 495425 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4200288 - -
          Illumina mouse-6 v1.0 expression beadchip 4200288 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004200288 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2700505 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2700505 - -
          mouse-6 v1.1 (Illumina) scl29744.12_13-S - -
          Illumina Sentrix mouse-6.v1 bead array 4200288 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl29744.12_13-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4200288 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2700505 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2831283 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2700505 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2831283 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl29744.12_13-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline