Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00512969_m1
          See other TMEM101 GE Assays ›
          Gene Symbol:
          Tmem101
          Gene Name:
          transmembrane protein 101
          Gene Aliases:
          2610511E22Rik
          Chromosome Location:
          Chr.11: 102152547 - 102156407 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00512969_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem101
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029649.2 NP_083925.2 3 - 4 529 65
          GenBank mRNA AK012106.1 - 3 - 4 483 65
          AK041154.1 - 3 - 4 488 65
          AK049378.1 - 3 - 4 511 65
          AK140260.1 - 3 - 4 530 65
          BC011109.1 - 3 - 4 499 2654288 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100003827.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem101

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          signal transducer activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28077836 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28077839 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28077838 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28077837 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P365656 AK049378.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P365656 AK140260.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P365656 BC011109.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P365656 NM_029649.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P365656 AK012106.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P365656 AK049378.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P365656 AK140260.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P365656 BC011109.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P365656 NM_029649.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P365656 AK012106.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 736255 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 486501 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P109198 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P365656 AK049378.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P365656 AK140260.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P365656 BC011109.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P365656 NM_029649.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P365656 AK012106.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P365656 AK049378.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P365656 AK140260.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P365656 BC011109.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P365656 NM_029649.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P365656 AK012106.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28077829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28077854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28077825 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28077830 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28077861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28077837 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28077840 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28077835 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28077832 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28077828 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28077823 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28077834 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28077851 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28077836 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28077845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28077857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28077824 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28077853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28077839 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28077831 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28077855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28077844 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28077859 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28077852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28077849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28077826 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28077842 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P109198 - -
          Whole Mouse Genome G4122A A_51_P365656 AK049378.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P365656 AK140260.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P365656 BC011109.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P365656 NM_029649.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P365656 AK012106.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P365656 AK049378.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P365656 AK140260.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P365656 BC011109.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P365656 NM_029649.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P365656 AK012106.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128581 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P109198 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09456966 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09456965 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09180965 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09456962 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28077836 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28077837 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28077839 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28077838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28077772 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28077837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09196451 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28077824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09077034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28077834 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28077805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28077832 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28077866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09077029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28077843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28077831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28077840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28077825 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28077821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28077842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28077849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28077835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09679080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28077812 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28077873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28077851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09627083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09174186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28077855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09679073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28077859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28077833 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28077815 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28077818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28077810 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28077868 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09456966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28077817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28077820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09664348 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28077860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09707104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28077841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28077864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28077857 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28077871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28077814 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09276612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28077852 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09707103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28077865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28077806 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10391571 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391568 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391570 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391569 - -
          Affymetrix HT MG-430 PM Array Plate 1451315_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10391567 - -
          Mouse Genome 430 2.0 Array 1451315_PM_at - -
          Mouse Genome 430 2.0 Array 1451315_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76547_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76547_at - -
          Affymetrix Mouse Clariom S Array PSR1100035476.mm.1 NM_029649.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100035476.mm.1 NM_029649.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100035476.mm.1 TC1100003827.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100035474.mm.1 NM_029649.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100035474.mm.1 NM_029649.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100035474.mm.1 TC1100003827.mm.1 MTA TC
          Mouse Expression Set 430 1451315_PM_at - -
          Mouse Expression Set 430 1451315_at - -
          Mouse Genome 430A 2.0 Array 1451315_PM_at - -
          Mouse Genome 430A 2.0 Array 1451315_at - -
          Affymetrix HT Mouse Genome 430A Array 1451315_at - -
          Affymetrix HT Mouse Genome 430A Array 1451315_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 917946 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 50575 - -
          Illumina mouse-6 v1.0 expression beadchip 50575 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000050575 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1245872 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1245872 - -
          mouse-6 v1.1 (Illumina) scl39518.4_244-S - -
          Illumina Sentrix mouse-6.v1 bead array 50575 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl39518.4_244-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 50575 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1245872 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1245872 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl39518.4_244-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline