Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00515567_m1
          See other CRYAB GE Assays ›
          Gene Symbol:
          Cryab
          Gene Name:
          crystallin, alpha B
          Gene Aliases:
          Crya-2, Crya2, HspB5, P23
          Chromosome Location:
          Chr.9: 50751325 - 50756636 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00515567_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cryab
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001289782.1 NP_001276711.1 3 - 4 772 59
          NM_001289784.1 NP_001276713.1 3 - 4 540 59
          NM_001289785.1 NP_001276714.1 3 - 4 529 59
          NM_009964.3 NP_034094.1 2 - 3 630 59
          GenBank mRNA AK146556.1 - 3 - 4 508 59
          AK168999.1 - 2 - 3 369 59
          BC010768.1 - 3 - 4 491 4238233 59
          BC094033.1 - 3 - 4 772 4934471 59
          CT010341.1 - 2 - 3 324 59
          M63170.1 - 2 - 3 324 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900000605.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cryab

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          lens development in camera-type eye
          protein folding
          tubulin complex assembly
          muscle organ development
          negative regulation of gene expression
          regulation of cell death
          negative regulation of cell growth
          microtubule polymerization or depolymerization
          negative regulation of intracellular transport
          response to hydrogen peroxide
          camera-type eye development
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          protein homooligomerization
          stress-activated MAPK cascade
          apoptotic process involved in morphogenesis
          cellular response to gamma radiation
          negative regulation of reactive oxygen species metabolic process
          structural constituent of eye lens
          protein binding
          microtubule binding
          cytoskeletal protein binding
          identical protein binding
          protein homodimerization activity
          metal ion binding
          unfolded protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2007656 BC010768.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2007656 BC094033.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2007656 CT010341.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2007656 M63170.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2007656 AK168999.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964408 BC010768.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964408 BC094033.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964408 M63170.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964408 AK168999.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964408 NM_009964.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 618599 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 106175 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 224417 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106503 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2007656 BC010768.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2007656 BC094033.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2007656 CT010341.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2007656 M63170.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2007656 AK168999.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964408 BC010768.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964408 BC094033.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964408 M63170.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964408 AK168999.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964408 NM_009964.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P504423 BC094033.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P504423 NM_009964.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P106482 BC010768.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P106482 BC094033.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P106482 CT010341.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P106482 M63170.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P106482 AK168999.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505447 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505469 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505462 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505446 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505465 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505463 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505439 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505460 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505435 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505444 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505450 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505454 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505451 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505464 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505458 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505461 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505456 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505452 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505437 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505467 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26505466 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106503 - -
          Whole Mouse Genome G4122A A_52_P106482 BC010768.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P106482 BC094033.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P106482 CT010341.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P106482 M63170.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P106482 AK168999.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P504423 BC094033.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P504423 NM_009964.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107220 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106503 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07804153 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07804134 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07804157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26505517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08030036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505460 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505466 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505437 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505472 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505446 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08154110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505439 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505461 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07773193 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505475 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505429 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07773192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505455 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505444 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505451 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505463 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07804171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07804163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505450 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26505452 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07773178 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10585215 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585218 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585220 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593419 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585219 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585216 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585222 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585223 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585217 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585221 - -
          Affymetrix HT MG-430 PM Array Plate 1434369_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1435837_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1416455_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10593418 - -
          Affymetrix Mouse Gene 1.1 ST Array 10585214 - -
          Mouse Genome 430 2.0 Array 1435837_at - -
          Mouse Genome 430 2.0 Array 1416455_PM_a_at - -
          Mouse Genome 430 2.0 Array 1434369_a_at - -
          Mouse Genome 430 2.0 Array 1435837_PM_at - -
          Mouse Genome 430 2.0 Array 1416455_a_at - -
          Mouse Genome 430 2.0 Array 1434369_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12955_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12955_at - -
          Affymetrix Mouse Clariom S Array PSR0900004300.mm.1 NM_001289782.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900004300.mm.1 NM_001289784.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900004300.mm.1 NM_001289785.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900004300.mm.1 NM_009964.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900004300.mm.1 TC0900000605.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900004297.mm.1 NM_001289782.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900004297.mm.1 NM_009964.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900004297.mm.1 TC0900000605.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900004291.mm.1 NM_001289782.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900004291.mm.1 TC0900000605.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900004299.mm.1 NM_001289782.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900004299.mm.1 NM_001289784.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900004299.mm.1 NM_001289785.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900004299.mm.1 NM_009964.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900004299.mm.1 TC0900000605.mm.1 MTA TC
          Mouse Expression Set 430 1416455_PM_a_at - -
          Mouse Expression Set 430 1416455_a_at - -
          Mouse Expression Set 430 1434369_a_at - -
          Mouse Expression Set 430 1435837_PM_at - -
          Mouse Expression Set 430 1434369_PM_a_at - -
          Mouse Expression Set 430 1435837_at - -
          Murine Genome U74v2 Set 93279_at - -
          Murine Genome U74v2 Set 93279_PM_at - -
          Murine Genome U74v2 Set 162308_f_at - -
          Murine Genome U74v2 Set 162308_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1416455_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1416455_a_at - -
          Mouse Genome 430A 2.0 Array 1435837_at - -
          Mouse Genome 430A 2.0 Array 1434369_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1435837_PM_at - -
          Mouse Genome 430A 2.0 Array 1434369_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1416455_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1416455_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1434369_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1434369_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1435837_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1435837_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 913499 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4810619 - -
          Illumina mouse-6 v1.0 expression beadchip 4810619 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004810619 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2687797 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2687797 - -
          mouse-6 v1.1 (Illumina) scl36966.5.1_28-S - -
          Illumina Sentrix mouse-6.v1 bead array 4810619 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl36966.5.1_28-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4810619 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2840213 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2840213 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl36966.5.1_28-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline