Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00515772_m1
          See other FHL1 GE Assays ›
          Gene Symbol:
          Fhl1
          Gene Name:
          four and a half LIM domains 1
          Gene Aliases:
          FHL-1, KyoT, RAM14-1, SLIM, SLIM-1
          Chromosome Location:
          Chr.X: 56731720 - 56793346 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00515772_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fhl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001077361.1 NP_001070829.1 2 - 3 271 66
          NM_010211.3 NP_034341.2 2 - 3 167 66
          GenBank mRNA AK128904.1 - 2 - 3 141 66
          AK158966.1 - 2 - 3 199 66
          U41739.1 - 1 - 2 58 66
          U77039.1 - 2 - 3 111 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00000525.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the NM_001077361.1,NM_010211.3,U41739.1,U77039.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fhl1

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of membrane depolarization
          multicellular organism development
          negative regulation of G2/M transition of mitotic cell cycle
          cell differentiation
          negative regulation of cell growth
          positive regulation of potassium ion transport
          regulation of potassium ion transmembrane transporter activity
          negative regulation of G1/S transition of mitotic cell cycle
          zinc ion binding
          ion channel binding
          metal ion binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0X00003457.mm.1 NM_001077361.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003457.mm.1 NM_001077361.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003457.mm.1 TC0X00000525.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00003454.mm.1 NM_010211.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003454.mm.1 TC0X00000525.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00003459.mm.1 NM_001077361.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003459.mm.1 NM_001077361.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003459.mm.1 NM_010211.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003459.mm.1 TC0X00000525.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00003471.mm.1 NM_001077361.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003471.mm.1 NM_001077361.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003471.mm.1 NM_001077362.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003471.mm.1 NM_001287800.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003471.mm.1 NM_010211.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003471.mm.1 TC0X00000525.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00003453.mm.1 NM_010211.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003453.mm.1 TC0X00000525.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00003458.mm.1 NM_001077361.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003458.mm.1 NM_001077361.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003458.mm.1 TC0X00000525.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 930146 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-fz59j:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0