Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00516330_m1
          See other AZGP1 GE Assays ›
          Gene Symbol:
          Azgp1
          Gene Name:
          alpha-2-glycoprotein 1, zinc
          Gene Aliases:
          Zag
          Chromosome Location:
          Chr.5: 137981521 - 137990233 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00516330_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Azgp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013478.2 NP_038506.2 2 - 3 365 83
          GenBank mRNA AK005051.1 - 2 - 3 351 83
          AY248694.1 - 2 - 3 326 83
          BC061646.1 - 2 - 3 347 5100216 83
          D21059.1 - 2 - 3 326 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500001638.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Azgp1

          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          detection of chemical stimulus involved in sensory perception of bitter taste
          antigen processing and presentation of peptide antigen via MHC class I
          immune response
          cell adhesion
          antigen processing and presentation
          regulation of multicellular organism growth
          glycoprotein binding
          antigen binding
          peptide antigen binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P456721 BC061646.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P456721 D21059.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P456721 BC061646.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P456721 D21059.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 169516 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 500634 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 84279 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 54908 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 265332 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 4788 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P167972 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P456721 BC061646.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P456721 D21059.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P456721 BC061646.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P456721 D21059.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P456721 BC061646.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P456721 D21059.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P456721 BC061646.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P456721 D21059.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24128456 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24128467 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24128465 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24128454 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24128440 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24128448 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24128464 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24128435 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24128444 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24128442 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24128441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24128446 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24128459 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24128462 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24128452 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24128455 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24128451 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24128438 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24128436 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24128433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24128450 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P167972 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05762020 - -
          Whole Mouse Genome G4122A A_51_P456721 BC061646.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P456721 D21059.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P456721 BC061646.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P456721 D21059.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108120 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105846 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P167972 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04383997 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04244621 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04244620 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04384000 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04383992 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04383998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128478 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04238049 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24128435 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04140264 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24128452 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24128465 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24128475 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24128482 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04160419 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24128473 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04393965 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24128431 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04393963 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04665605 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04383992 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04319188 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24128477 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24128425 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04319189 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24128436 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24128480 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24128472 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04113171 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04238053 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04143078 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04990055 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04244621 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24128441 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24128454 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04140263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04238050 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04390340 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24128440 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04393967 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04766754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04384000 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04990053 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24128479 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24128455 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24128446 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24128438 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24128467 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24128478 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10526717 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526716 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526713 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526714 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526715 - -
          Affymetrix HT MG-430 PM Array Plate 1417776_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10526712 - -
          Mouse Genome 430 2.0 Array 1417776_PM_at - -
          Mouse Genome 430 2.0 Array 1417776_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12007_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12007_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500014971.mm.1 TC0500001638.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500014971.mm.1 NM_013478.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014971.mm.1 NM_013478.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014972.mm.1 TC0500001638.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500014972.mm.1 NM_013478.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014972.mm.1 NM_013478.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014973.mm.1 TC0500001638.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500014973.mm.1 NM_013478.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014973.mm.1 NM_013478.2 Refseq
          Mouse Expression Set 430 1417776_PM_at - -
          Mouse Expression Set 430 1417776_at - -
          Murine Genome U74v2 Set 96867_PM_at - -
          Murine Genome U74v2 Set 96867_at - -
          Mouse Genome 430A 2.0 Array 1417776_at - -
          Mouse Genome 430A 2.0 Array 1417776_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417776_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417776_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 621177 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1340411 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001340411 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2725198 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2725198 - -
          mouse-6 v1.1 (Illumina) scl27087.3.5_17-S - -
          Illumina Sentrix mouse-6.v1 bead array 1340411 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl27087.3.5_17-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1340411 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2725198 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2837080 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2837080 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2725198 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline