Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00517944_m1
          See other CIAO1 GE Assays ›
          Gene Symbol:
          Ciao1
          Gene Name:
          cytosolic iron-sulfur protein assembly 1
          Gene Aliases:
          AW210570, Wdr39
          Chromosome Location:
          Chr.2: 127240938 - 127247816 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00517944_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ciao1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025296.4 NP_079572.2 6 - 7 980 76
          GenBank mRNA AK002314.1 - 6 - 7 925 76
          AK028376.1 - 6 - 7 907 76
          BC004089.1 - 6 - 7 901 3593736 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200004550.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ciao1

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription from RNA polymerase II promoter
          chromosome segregation
          iron-sulfur cluster assembly
          transcription factor activity, sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21612303 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21612301 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21612300 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21612304 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21612302 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P585699 AK028376.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P585699 AK080259.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P585699 NM_025296.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P585699 AK028376.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P585699 AK080259.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P585699 NM_025296.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P118195 AK028376.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P118195 BC004089.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P118195 NM_025296.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P118195 AK002314.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P118195 AK004129.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 184020 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 889270 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 824028 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 330172 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 10857 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 298706 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 263072 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 310097 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 582396 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 71729 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 145603 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 42104 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 45436 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 55913 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100253 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P585699 AK028376.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P585699 AK080259.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P585699 NM_025296.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P585699 AK028376.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P585699 AK080259.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P585699 NM_025296.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P118195 AK028376.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P118195 BC004089.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P118195 NM_025296.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P118195 AK002314.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P118195 AK004129.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P124951 BC004089.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P124951 BU555826.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P124951 NM_025296.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P124951 AK002314.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612270 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612290 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612302 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612269 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612274 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612287 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612277 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612304 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612280 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612283 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612281 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612284 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612285 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612293 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612295 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612267 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04684418 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100253 - -
          Whole Mouse Genome G4122A A_51_P124951 BC004089.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P124951 BU555826.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P124951 NM_025296.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P124951 AK002314.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P585699 AK028376.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P585699 AK080259.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P585699 NM_025296.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P585699 AK028376.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P585699 AK080259.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P585699 NM_025296.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111179 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118195 AK028376.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118195 BC004089.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118195 NM_025296.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118195 AK002314.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118195 AK004129.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100253 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02093269 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02093263 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02093274 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21612302 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21612304 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21612303 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21612300 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21612301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612300 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01956635 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612274 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612269 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01167783 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612264 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612283 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01767475 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02093274 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612259 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612270 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02093277 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612260 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01953183 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612273 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01167784 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612280 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01956634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612302 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612267 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612278 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612298 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612281 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01167781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612277 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612266 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612275 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01767472 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10487377 - -
          Affymetrix Mouse Gene 1.0 ST Array 10487378 - -
          Affymetrix Mouse Gene 1.0 ST Array 10487379 - -
          Affymetrix Mouse Gene 1.0 ST Array 10487372 - -
          Affymetrix Mouse Gene 1.0 ST Array 10487375 - -
          Affymetrix Mouse Gene 1.0 ST Array 10487374 - -
          Affymetrix Mouse Gene 1.0 ST Array 10487373 - -
          Affymetrix Mouse Gene 1.0 ST Array 10487376 - -
          Affymetrix HT MG-430 PM Array Plate 1450953_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10487371 - -
          Mouse Genome 430 2.0 Array 1450953_at - -
          Mouse Genome 430 2.0 Array 1450953_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 26371_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 26371_at - -
          Affymetrix Mouse Clariom S Array PSR0200038972.mm.1 NM_025296.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038972.mm.1 NM_025296.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038972.mm.1 TC0200004550.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200038973.mm.1 NM_025296.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038973.mm.1 NM_025296.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038973.mm.1 TC0200004550.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200038974.mm.1 NM_025296.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038974.mm.1 NM_025296.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038974.mm.1 TC0200004550.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200038976.mm.1 NM_025296.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038976.mm.1 NM_025296.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038976.mm.1 TC0200004550.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200038971.mm.1 NM_025296.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038971.mm.1 NM_025296.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038971.mm.1 TC0200004550.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200038977.mm.1 NM_025296.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038977.mm.1 NM_025296.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038977.mm.1 TC0200004550.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200038975.mm.1 NM_025296.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038975.mm.1 NM_025296.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038975.mm.1 TC0200004550.mm.1 MTA TC
          Mouse Expression Set 430 1450953_PM_at - -
          Mouse Expression Set 430 1450953_at - -
          Murine Genome U74v2 Set 100072_PM_at - -
          Murine Genome U74v2 Set 100072_at - -
          Murine Genome U74v2 Set 134069_PM_at - -
          Murine Genome U74v2 Set 134069_at - -
          Mouse Genome 430A 2.0 Array 1450953_PM_at - -
          Mouse Genome 430A 2.0 Array 1450953_at - -
          Affymetrix HT Mouse Genome 430A Array 1450953_at - -
          Affymetrix HT Mouse Genome 430A Array 1450953_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 711751 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5390097 - -
          Illumina mouse-6 v1.0 expression beadchip 5390097 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005390097 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2650944 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2650944 - -
          mouse-6 v1.1 (Illumina) scl18709.8_168-S - -
          Illumina Sentrix mouse-6.v1 bead array 5390097 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl18709.8_168-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5390097 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1218843 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1218843 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl18709.8_168-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline