Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00518984_m1
          See other IL23A GE Assays ›
          Gene Symbol:
          Il23a
          Gene Name:
          interleukin 23, alpha subunit p19
          Gene Aliases:
          IL-23, p19
          Chromosome Location:
          Chr.10: 128296140 - 128298084 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00518984_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Il23a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031252.2 NP_112542.1 2 - 3 377 61
          GenBank mRNA AF301619.1 - 2 - 3 377 61
          AK043894.1 - 1264 61
          BC019953.1 - 2 - 3 361 3708687 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000003135.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Il23a

          Panther Classification:

          Molecular Function -

          cytokine interleukin superfamily intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of T cell mediated cytotoxicity
          positive regulation of defense response to virus by host
          immune system process
          positive regulation of T-helper 1 type immune response
          inflammatory response
          immune response
          positive regulation of activation of JAK2 kinase activity
          negative regulation of interleukin-10 production
          positive regulation of granulocyte macrophage colony-stimulating factor production
          positive regulation of interferon-gamma production
          positive regulation of interleukin-10 production
          positive regulation of interleukin-12 production
          positive regulation of interleukin-17 production
          positive regulation of tumor necrosis factor production
          positive regulation of natural killer cell proliferation
          T cell proliferation
          positive regulation of T cell proliferation
          positive regulation of activated T cell proliferation
          regulation of tyrosine phosphorylation of Stat1 protein
          positive regulation of tyrosine phosphorylation of Stat3 protein
          positive regulation of tyrosine phosphorylation of Stat4 protein
          positive regulation of tyrosine phosphorylation of Stat5 protein
          positive regulation of memory T cell differentiation
          innate immune response
          positive regulation of osteoclast differentiation
          positive regulation of transcription from RNA polymerase II promoter
          tissue remodeling
          defense response to Gram-negative bacterium
          positive regulation of NK T cell activation
          positive regulation of NK T cell proliferation
          defense response to virus
          positive regulation of neutrophil chemotaxis
          positive regulation of T-helper 17 type immune response
          positive regulation of T-helper 17 cell lineage commitment
          cytokine activity
          protein binding
          interleukin-23 receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27540624 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27540625 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27540626 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P649817 AK043894.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P649817 BC019953.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P649817 NM_031252.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P649817 AF301619.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P649817 AK043894.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P649817 BC019953.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P649817 NM_031252.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P649817 AF301619.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 776448 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 528038 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P104880 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P649817 AK043894.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P649817 BC019953.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P649817 NM_031252.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P649817 AF301619.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P649817 AK043894.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P649817 BC019953.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P649817 NM_031252.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P649817 AF301619.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27540604 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27540602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27540592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27540597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27540595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27540599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27540582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27540579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27540581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27540574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27540624 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27540611 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27540584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27540590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27540625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27540580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27540617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27540608 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27540610 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27540575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27540618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27540578 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27540588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27540606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27540593 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27540596 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27540615 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27540577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27540614 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27540620 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P104880 - -
          Whole Mouse Genome G4122A A_52_P649817 AK043894.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P649817 BC019953.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P649817 NM_031252.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P649817 AF301619.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P649817 AK043894.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P649817 BC019953.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P649817 NM_031252.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P649817 AF301619.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121017 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P104880 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08666687 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08666683 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08666682 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08473805 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08839082 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08839081 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08473808 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08839083 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08839098 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08473807 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27540625 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27540626 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27540624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27540602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27540578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08388630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08663104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08666686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27540584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27540602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27540575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27540582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27540577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27540581 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08666666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27540608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27540579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27540604 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08469414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08473807 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08473810 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27540593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27540609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27540595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27540569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27540603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27540588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27540580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27540598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27540592 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10373359 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373362 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373360 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373361 - -
          Affymetrix HT MG-430 PM Array Plate 1419529_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10373358 - -
          Mouse Genome 430 2.0 Array 1419529_PM_at - -
          Mouse Genome 430 2.0 Array 1419529_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 83430_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 83430_at - -
          Affymetrix Mouse Clariom S Array PSR1000023523.mm.1 NM_031252.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023523.mm.1 NM_031252.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023523.mm.1 TC1000003135.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000023522.mm.1 NM_031252.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023522.mm.1 NM_031252.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023522.mm.1 TC1000003135.mm.1 MTA TC
          Mouse Expression Set 430 1419529_at - -
          Mouse Expression Set 430 1419529_PM_at - -
          Mouse Genome 430A 2.0 Array 1419529_PM_at - -
          Mouse Genome 430A 2.0 Array 1419529_at - -
          Affymetrix HT Mouse Genome 430A Array 1419529_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419529_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 710568 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6290044 - -
          Illumina mouse-6 v1.0 expression beadchip 6290044 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006290044 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2671320 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2671320 - -
          mouse-6 v1.1 (Illumina) scl37367.3.1_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 6290044 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37367.3.1_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6290044 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2671320 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2799748 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2671320 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2799748 - -

          Back To Top
           

          Citations (1)

          CCR7 ligands are required for development of experimental autoimmune encephalomyelitis through generating IL-23-dependent Th17 cells.
          Authors
          Kuwabara T, Ishikawa F, Yasuda T, Aritomi K, Nakano H, Tanaka Y, Okada Y, Lipp M, Kakiuchi T
          Journal
          J Immunol - Page No 2513 (Sat Aug 15 00:00:00 EDT 2009)
          Volume (Year)
          183 (2009)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0