Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00518988_m1
          See other IFI44L GE Assays ›
          Gene Symbol:
          Ifi44l
          Gene Name:
          interferon-induced protein 44 like
          Gene Aliases:
          H-28, H28, H28-1, NS1178
          Chromosome Location:
          Chr.3: 151758737 - 151762891 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00518988_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ifi44l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031367.1 NP_112735.1 1234 143
          GenBank mRNA AF336221.1 - 1234 143

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ifi44l

          Gene Ontology Categories:

          Function(s)

          immune response

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P327657 NM_031367.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P327657 AF336221.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P327657 NM_031367.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P327657 AF336221.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 73349 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 818267 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P165140 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139319 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P327657 NM_031367.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P327657 AF336221.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P327657 NM_031367.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P327657 AF336221.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22646997 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22647002 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22646995 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22647005 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22647001 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22646986 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22646980 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22646984 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22646971 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22646990 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22646978 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22646994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22647000 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22646973 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22646977 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22646982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22646988 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22647004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22646993 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22646976 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P165140 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139319 - -
          Whole Mouse Genome G4122A A_51_P327657 NM_031367.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P327657 AF336221.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P327657 NM_031367.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P327657 AF336221.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P327651 NM_031367.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P327651 AF336221.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116166 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P165140 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139319 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02438986 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02410928 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02438985 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02438981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22646927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22647014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22646997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22646921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22646912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22646976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22646940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22646994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22646980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22646984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22646913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22647008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22646993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22646995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22646965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22646907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22647004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22646903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22646915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22646942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22646910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22646937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22646932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22646969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22646939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22646978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22646988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22647022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22646923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22646967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22646961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22647021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22647017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22646986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22646972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22647009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22646928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22647001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22647011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22647002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22646982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22646909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22647007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22646925 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22647002 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22646988 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22646978 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22646997 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02657665 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22646971 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22646995 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22646994 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22646998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02410928 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22646972 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02438984 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22646984 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22646961 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22646991 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02503130 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22646980 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02657654 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22646967 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03011861 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22646969 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22647014 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22646960 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22646965 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22646986 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22647007 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22646982 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22646976 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22647004 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22647001 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10502802 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502804 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502803 - -
          Affymetrix HT MG-430 PM Array Plate 1425917_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1421596_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1450301_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10502801 - -
          Mouse Genome 430 2.0 Array 1421596_PM_s_at - -
          Mouse Genome 430 2.0 Array 1425917_PM_at - -
          Mouse Genome 430 2.0 Array 1421596_s_at - -
          Mouse Genome 430 2.0 Array 1450301_PM_at - -
          Mouse Genome 430 2.0 Array 1425917_at - -
          Mouse Genome 430 2.0 Array 1450301_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15061_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15061_at - -
          Mouse Expression Set 430 1450301_PM_at - -
          Mouse Expression Set 430 1421596_s_at - -
          Mouse Expression Set 430 1421596_PM_s_at - -
          Mouse Expression Set 430 1425917_PM_at - -
          Mouse Expression Set 430 1425917_at - -
          Mouse Expression Set 430 1450301_at - -
          Mouse Genome 430A 2.0 Array 1425917_PM_at - -
          Mouse Genome 430A 2.0 Array 1450301_at - -
          Mouse Genome 430A 2.0 Array 1421596_s_at - -
          Mouse Genome 430A 2.0 Array 1421596_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1425917_at - -
          Mouse Genome 430A 2.0 Array 1450301_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425917_at - -
          Affymetrix HT Mouse Genome 430A Array 1425917_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421596_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1421596_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1450301_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450301_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 7100563 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007100563 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2645225 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2645225 - -
          mouse-6 v1.1 (Illumina) scl015061.6_43-S - -
          Illumina Sentrix mouse-6.v1 bead array 7100563 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl015061.6_43-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7100563 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2645225 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2645225 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline