Search
Search

Otud5| Gene Symbol: Otud5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290536.1 | NP_001277465.1 | 7 - 8 | 1748 | 79 | |
| NM_001290537.1 | NP_001277466.1 | 8 - 9 | 1368 | 79 | ||
| NM_138604.3 | NP_613070.2 | 7 - 8 | 1748 | 79 | ||
| GenBank mRNA | AF229642.1 | - | 7 - 8 | 889 | 79 | |
| AK009620.1 | - | 2 - 3 | 264 | 79 | ||
| AK010750.1 | - | 8 - 9 | 1368 | 79 | ||
| AK149777.1 | - | 8 - 9 | 1369 | 79 | ||
| AK155131.1 | - | 7 - 8 | 1724 | 79 | ||
| AK167226.1 | - | 6 - 7 | 1061 | 79 | ||
| AK169921.1 | - | 9 - 10 | 1316 | 79 | ||
| AK171874.1 | - | 7 - 8 | 1489 | 79 | ||
| AK180926.1 | - | 1 - 2 | 196 | 79 | ||
| AK199214.1 | - | 1 - 2 | 196 | 79 | ||
| AK209520.1 | - | 1 - 2 | 196 | 79 | ||
| BC016529.1 | - | 7 - 8 | 1748 | 3594583 | 79 | |
| BC051111.1 | - | 7 - 8 | 1487 | 5253679 | 79 | |
Set Membership: |
|