Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00519781_m1
          See other DEFB9 GE Assays ›
          Gene Symbol:
          Defb9
          Gene Name:
          defensin beta 9
          Gene Aliases:
          BD-9
          Chromosome Location:
          Chr.8: 21881713 - 21885434 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00519781_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Defb9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_139219.2 NP_631965.1 1 - 2 105 68
          GenBank mRNA AJ437647.1 - 1 - 2 92 68
          BC109177.2 - 1 - 2 88 40049218 68
          BM936051.1 - 1 - 2 134 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001855.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Defb9

          Panther Classification:

          Molecular Function -

          antimicrobial response protein defense/immunity protein

          Gene Ontology Categories:

          Function(s) Process(es)

          defense response
          biological_process
          defense response to bacterium
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957729 AJ437647.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957729 BC109177.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957729 BM936051.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957729 NM_139219.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 774558 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 236974 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 48687 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P145356 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957729 AJ437647.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957729 BC109177.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957729 BM936051.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957729 NM_139219.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25708100 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25708118 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25708106 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25708097 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25708111 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25708101 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25708120 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25708123 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25708096 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25708107 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25708114 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25708115 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P145356 - -
          Whole Mouse Genome G4122A A_51_P124362 AJ437647.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P124362 BC109177.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P124362 BM936051.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P124362 NM_139219.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121192 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113509 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P145356 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06698261 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06698262 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06698265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07056992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708137 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06698262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06931817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07179284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06940242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708088 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07179287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07179290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708115 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10577442 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577443 - -
          Affymetrix HT MG-430 PM Array Plate 1427612_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10577441 - -
          Mouse Genome 430 2.0 Array 1427612_PM_at - -
          Mouse Genome 430 2.0 Array 1427612_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 246079_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 246079_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0800014621.mm.1 NM_139219.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014621.mm.1 NM_139219.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014621.mm.1 TC0800001855.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800014622.mm.1 NM_139219.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014622.mm.1 NM_139219.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014622.mm.1 TC0800001855.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800014623.mm.1 NM_139219.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014623.mm.1 NM_139219.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014623.mm.1 TC0800001855.mm.1 MTA TC
          Mouse Expression Set 430 1427612_PM_at - -
          Mouse Expression Set 430 1427612_at - -
          Mouse Genome 430A 2.0 Array 1427612_PM_at - -
          Mouse Genome 430A 2.0 Array 1427612_at - -
          Affymetrix HT Mouse Genome 430A Array 1427612_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427612_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 773180 - -
          ABI Mouse Genome Survey Microarray 889736 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6590377 - -
          Illumina mouse-6 v1.0 expression beadchip 6590377 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006590377 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2680517 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2680517 - -
          mouse-6 v1.1 (Illumina) scl35032.2.1_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 6590377 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl35032.2.1_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6590377 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2680517 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2680517 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0