Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00519870_m1
          See other CAR9 GE Assays ›
          Gene Symbol:
          Car9
          Gene Name:
          carbonic anhydrase 9
          Gene Aliases:
          CAIX, Ca9, MN/CA9
          Chromosome Location:
          Chr.4: 43507026 - 43513810 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00519870_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Car9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_139305.2 NP_647466.2 5 - 6 808 70
          GenBank mRNA AB086322.1 - 5 - 6 810 70
          AJ245857.1 - 5 - 6 810 70
          AK136579.1 - 5 - 6 809 70
          AK137719.1 - 5 - 6 810 70
          AK166403.1 - 2 - 3 237 70
          BC120544.1 - 5 - 6 801 40129467 70
          BC120546.1 - 5 - 6 801 40129469 70
          EF122497.1 - 5 - 6 810 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400000438.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Car9

          Gene Ontology Categories:

          Function(s) Process(es)

          morphogenesis of an epithelium
          one-carbon metabolic process
          secretion
          carbonate dehydratase activity
          zinc ion binding
          lyase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2092750 EF122497.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2092750 BC057148.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2092750 AB086322.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2092750 AJ245857.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2092750 AK136579.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 517344 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 526115 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 9918 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 364561 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P119529 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2092750 EF122497.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2092750 BC057148.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2092750 AB086322.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2092750 AJ245857.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2092750 AK136579.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P610923 BC120546.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P610923 EF122497.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P610923 AK137719.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P610923 AK166403.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P610923 BC120544.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872088 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872090 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872087 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872078 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872084 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872073 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872079 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872076 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872081 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872080 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05217231 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P119529 - -
          Whole Mouse Genome G4122A A_52_P610923 BC120546.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P610923 EF122497.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P610923 AK137719.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P610923 AK166403.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P610923 BC120544.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134932 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P119529 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03950029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872165 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03743729 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03857565 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872092 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872121 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03950037 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872042 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03754468 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03522959 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872079 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872066 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872074 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03754476 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872064 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872094 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03600680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872105 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872050 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872052 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872118 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872054 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03754480 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03310128 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872102 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03600693 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03600674 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872084 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03279631 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872110 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872063 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872099 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872078 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872115 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872056 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872043 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872081 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03754459 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03310134 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872111 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872067 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03950034 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872088 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872100 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872080 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03754472 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03522962 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872120 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872122 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872095 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10504345 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504341 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504338 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504346 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504339 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504343 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504348 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504344 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504347 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504340 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504342 - -
          Affymetrix HT MG-430 PM Array Plate 1427013_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10504337 - -
          Mouse Genome 430 2.0 Array 1427013_at - -
          Mouse Genome 430 2.0 Array 1427013_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 230099_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 230099_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400003004.mm.1 NM_139305.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003004.mm.1 NM_139305.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003004.mm.1 TC0400000438.mm.1 MTA TC
          Mouse Expression Set 430 1427013_PM_at - -
          Mouse Expression Set 430 1427013_at - -
          Murine Genome U74v2 Set 167049_at - -
          Murine Genome U74v2 Set 167049_PM_at - -
          Mouse Genome 430A 2.0 Array 1427013_at - -
          Mouse Genome 430A 2.0 Array 1427013_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427013_at - -
          Affymetrix HT Mouse Genome 430A Array 1427013_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 670057 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2230113 - -
          MouseWG-6 v2 Expression BeadChip 2340128 - -
          Illumina mouse-6 v1.0 expression beadchip 2230113 - -
          Illumina mouse-6 v1.0 expression beadchip 2340128 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002340128 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002230113 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2730999 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1244145 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1244145 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2730999 - -
          mouse-6 v1.1 (Illumina) scl25514.12.16_2-S - -
          mouse-6 v1.1 (Illumina) scl0002701.1_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 2230113 - -
          Illumina Sentrix mouse-6.v1 bead array 2340128 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl25514.12.16_2-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0002701.1_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2230113 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2340128 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1244145 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2730999 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2874084 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2730999 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1244145 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2874084 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-knt4q:80/100.66.75.14:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0