Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00521145_m1
          See other SPG11 GE Assays ›
          Gene Symbol:
          Spg11
          Gene Name:
          spastic paraplegia 11
          Gene Aliases:
          6030465E24Rik, A330015I11, C530005A01Rik
          Chromosome Location:
          Chr.2: 122053526 - 122118415 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00521145_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spg11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145531.2 NP_663506.2 35 - 36 6567 118
          GenBank mRNA AK031637.1 - 12 - 13 2530 118
          AK083468.1 - 4 - 5 413 118
          AK129453.1 - 22 - 23 4001 118
          BC019404.1 - 6 - 7 757 118

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200004502.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Spg11

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P289414 BC019404.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P289414 BC019404.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 728437 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 571992 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 289053 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 339926 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 654966 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 839415 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 413942 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 168426 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 799795 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 110968 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 475510 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 727395 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 944403 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 645854 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 901028 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 197166 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 882951 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 556209 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 284535 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 656647 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 216578 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 895913 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 490101 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 731556 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 41868 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 711987 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 11013 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 524288 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 401645 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 219536 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 442832 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 15918 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 88514 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 355960 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 947175 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 413157 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 971194 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 694957 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 960959 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 148720 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 14878 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 94586 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 112394 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 109065 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 44243 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P170827 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P167886 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154015 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125634 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P289414 BC019404.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P289414 BC019404.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1970695 AK139974.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04672375 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P170827 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154015 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125634 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04672499 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04672402 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P167886 - -
          Whole Mouse Genome G4122A A_52_P150775 AK147500.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P150775 AK039286.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P150775 NM_145531.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P482578 AK147500.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P482578 AK049617.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P482578 AK049236.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P482578 AK039286.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P482578 NM_145531.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P409482 AK049617.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154015 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P167886 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P170827 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01742751 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21584379 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21584377 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01167627 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21584376 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21584381 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01167618 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21584387 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01167624 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10486913 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486919 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486904 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486909 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486914 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486901 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486916 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486925 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486917 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486918 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486907 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486908 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486905 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486927 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486920 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486923 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486930 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486903 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486934 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486915 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486912 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486900 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486931 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486924 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486933 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486926 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486910 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486922 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486929 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486928 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486911 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486921 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486899 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486932 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486902 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486906 - -
          Affymetrix HT MG-430 PM Array Plate 1426451_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1445021_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10486898 - -
          Mouse Genome 430 2.0 Array 1426451_at - -
          Mouse Genome 430 2.0 Array 1445021_at - -
          Mouse Genome 430 2.0 Array 1445021_PM_at - -
          Mouse Genome 430 2.0 Array 1426451_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200038189.mm.1 NM_145531.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038189.mm.1 NM_145531.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038189.mm.1 TC0200004502.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200038190.mm.1 NM_145531.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038190.mm.1 NM_145531.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038190.mm.1 TC0200004502.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200038186.mm.1 NM_145531.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038186.mm.1 NM_145531.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038186.mm.1 TC0200004502.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200038188.mm.1 NM_145531.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038188.mm.1 NM_145531.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038188.mm.1 TC0200004502.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200038187.mm.1 NM_145531.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038187.mm.1 NM_145531.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038187.mm.1 TC0200004502.mm.1 MTA TC
          Mouse Expression Set 430 1426451_PM_at - -
          Mouse Expression Set 430 1426451_at - -
          Mouse Expression Set 430 1445021_at - -
          Mouse Expression Set 430 1445021_PM_at - -
          Mouse Genome 430A 2.0 Array 1426451_PM_at - -
          Mouse Genome 430A 2.0 Array 1426451_at - -
          Affymetrix HT Mouse Genome 430A Array 1426451_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1426451_at - -
          Affymetrix HT Mouse Genome 430B Array 1445021_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1445021_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 662423 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105550170 - -
          MouseWG-6 v2 Expression BeadChip 106840609 - -
          Illumina mouse-6 v1.0 expression beadchip 106840609 - -
          Illumina mouse-6 v1.0 expression beadchip 105550170 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1214565 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1246794 - -
          Illumina Sentrix mouse-6.v1 bead array 106840609 - -
          Illumina Sentrix mouse-6.v1 bead array 105550170 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105550170 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106840609 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1246794 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1214565 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0