Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00523731_g1
          See other RHOV GE Assays ›
          Gene Symbol:
          Rhov
          Gene Name:
          ras homolog gene family, member V
          Gene Aliases:
          A030005A06Rik, Arhv
          Chromosome Location:
          Chr.2: 119269201 - 119278008 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00523731_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rhov
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145530.2 NP_663505.1 1 - 2 313 78
          GenBank mRNA AK036775.1 - 1 - 2 314 78
          AK037170.1 - 1 - 2 315 78
          BC021307.1 - 1 - 2 318 5003249 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200004427.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rhov

          Panther Classification:

          Molecular Function -

          small GTPase

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          small GTPase mediated signal transduction
          nucleotide binding
          GTP binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21568738 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21568741 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21568739 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P439584 AK037170.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P439584 AK036775.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P439584 NM_145530.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P439584 AK037170.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P439584 AK036775.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P439584 NM_145530.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 624943 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P123577 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P439584 AK037170.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P439584 AK036775.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P439584 NM_145530.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P439584 AK037170.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P439584 AK036775.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P439584 NM_145530.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P439584 AK037170.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P439584 AK036775.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P439584 NM_145530.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P439584 AK037170.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P439584 AK036775.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P439584 NM_145530.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568725 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568727 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568736 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568723 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568739 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568745 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568737 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568734 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568730 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568755 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568732 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568749 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568742 - -
          Whole Mouse Genome G4122A A_51_P439584 AK037170.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P439584 AK036775.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P439584 NM_145530.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P439584 AK037170.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P439584 AK036775.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P439584 NM_145530.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129384 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02085469 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02085488 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02083738 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21568739 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21568738 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21568741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568705 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568745 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568753 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568701 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568742 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568700 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01257789 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568695 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568682 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02280220 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568705 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568707 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01522810 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568714 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01274903 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568715 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568691 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568702 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568756 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02085483 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568769 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01824097 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568674 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01634482 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02085493 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02239221 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01577164 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568749 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01577166 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568772 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568712 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02085488 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568770 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568711 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568699 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568685 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568676 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568746 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01560469 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01634483 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568747 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01447971 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568720 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568732 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568696 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568761 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02085469 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568727 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10486199 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486200 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486198 - -
          Affymetrix HT MG-430 PM Array Plate 1424976_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10486197 - -
          Mouse Genome 430 2.0 Array 1424976_at - -
          Mouse Genome 430 2.0 Array 1424976_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 228543_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 228543_at - -
          Affymetrix Mouse Clariom S Array PSR0200036987.mm.1 NM_145530.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200036987.mm.1 NM_145530.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200036987.mm.1 TC0200004427.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200036986.mm.1 NM_145530.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200036986.mm.1 NM_145530.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200036986.mm.1 TC0200004427.mm.1 MTA TC
          Mouse Expression Set 430 1424976_PM_at - -
          Mouse Expression Set 430 1424976_at - -
          Murine Genome U74v2 Set 133045_at - -
          Murine Genome U74v2 Set 133045_PM_at - -
          Mouse Genome 430A 2.0 Array 1424976_at - -
          Mouse Genome 430A 2.0 Array 1424976_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424976_at - -
          Affymetrix HT Mouse Genome 430A Array 1424976_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 850496 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5860100 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005860100 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1244918 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1244918 - -
          mouse-6 v1.1 (Illumina) scl18807.3_31-S - -
          Illumina Sentrix mouse-6.v1 bead array 5860100 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl18807.3_31-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5860100 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2859528 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2859528 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline